Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide Sequence: MMTAESREAT GLSPQAAQEK DGIVIAKVEE EDEEDHMWGQ DSTLQDTPPP DPEIFRQRFR RFCYQNTFGP REALSRLKEL CHQWLRPEIN TKEQILELLV LEQFLSILPK ELQVWLQEYR PDSGEEAVTL LEDLELDLSG QQVPGQVHGP EMLARGMVPL DPVQESSSFD LHHEATQSHF KHSSRKPRLL QSRALPAAHI PAPPHEGSPR DQAMASALFT ADSQAMVKIE DMAVSLILEE WGCQNLARRN LSRDNRQENY GSAFPQGGEN RNENEESTSK AETSEDSASR GETTGRSQKE FGEKRDQEGK TGERQQKNPE EKTRKEKRDS GPAIGKDKKT ITGERGPREK GKGLGRSFSL SSNFTTPEEV PTGTKSHRCD ECGKCFTRSS SLIRHKIIHT GEKPYECSEC GKAFSLNSNL VLHQRIHTGE KPHECNECGK AFSHSSNLIL HQRIHSGEKP YECNECGKAF SQSSDLTKHQ RIHTGKAFNR NSYLILHRRI HTREKPYKCT KCGKAFTRSS TLTLHHRIHA RERASEYSPA SLDAFGAFLK SCV
The ZKSCAN1 gene encodes a transcriptional regulator of the KRAB. Transcriptional regulatory proteins containing tandemly repeated zinc finger domains are thought to be involved in both normal and abnormal cellular proliferation and differentiation. See ZNF91 (MIM 603971) for general information on zinc finger proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: unnamed protein product; Zinc finger protein 139; Zinc finger protein 36; zinc finger protein 36 (KOX 18); Zinc finger protein KOX18; Zinc finger protein with KRAB and SCAN domains 1; ZNF36 isoform
Gene Aliases: KOX18; PHZ-37; ZKSCAN1; ZNF139; ZNF36; ZSCAN33
UniProt ID: (Human) P17029
Entrez Gene ID: (Human) 7586
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support