Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide Sequence: GHSNHIALYA AVRALHSEGK LPKGCSVLTL QSVNVLRKYI SLLDLPLSLL HTQDVLFVLN SKEVAQAKKA MSCHRSQLLW FRRLYIIFSR YMRINSLSFL
This gene encodes an enzyme that catalyzes the second step of glycosylphosphatidylinositol (GPI) biosynthesis, which is the de-N-acetylation of N-acetylglucosaminylphosphatidylinositol (GlcNAc-PI). Study of a similar rat enzyme suggests that this protein localizes to the endoplasmic reticulum.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: N-acetylglucosaminyl-phosphatidylinositol de-N-acetylase; N-acetylglucosaminylphosphatidylinositol deacetylase; phosphatidylinositol glycan, class L; Phosphatidylinositol-glycan biosynthesis class L protein; PIG-L; unnamed protein product
Gene Aliases: CHIME; PIGL
UniProt ID: (Human) Q9Y2B2
Entrez Gene ID: (Human) 9487
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support