Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
In direct ELISAs, no cross-reactivity with recombinant human LMIR1, 2, 4, 5, 6, or recombinant mouse LMIR3 is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
CLM1 is an inhibitory receptor of the Ig superfamily expressed on myeloid cells. It mediates negative regulatory signals by recruiting SHP1 (PTPN6; MIM 176883) or SHIP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD300 antigen-like family member F; CD300f; CLM-1; CMRF35-like molecule 1; IgSF13; immune receptor; Immune receptor expressed on myeloid cells 1; immunoglobin superfamily member 13; Immunoglobulin superfamily member 13; inhibitory receptor IREM1; IREM-1; IREM-1 Sv1; IREM-1 Sv2; NK inhibitory receptor; NKIR; unnamed protein product
Gene Aliases: CD300F; CD300LF; CLM-1; CLM1; IGSF13; IREM-1; IREM1; LMIR3; NKIR; UNQ3105/PRO10111
UniProt ID: (Human) Q8TDQ1
Entrez Gene ID: (Human) 146722
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support