Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
TNS4 links the focal adhesion and cell adhesion signaling to the actin cytoskeleton. It may be involved in cell survival and migration. The protein may be involved in cell migration, cartilage development and in linking signal transduction pathways to the cytoskeleton and promote apoptosis, via its cleavage by caspase-3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C terminal tensin like; c-ten; C-terminal tensin-like protein; cten; Tensin-4; unnamed protein product
Gene Aliases: CTEN; PP14434; TNS4
UniProt ID: (Human) Q8IZW8
Entrez Gene ID: (Human) 84951
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support