Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide Sequence: GDLLGEGPRH PLQPRERLYR AQLEVKVASE QTEKLLNKVL GSEPAPVSAE TLLSQAVEQL RQATQVLQEM RDLGELSQEA PGLREKRKEL VTLYRRSAP
Pleckstrin homology domain-containing proteins family O member 2 (PLEKHO2)'s function remains largely unknown. Genetic polymorphisms of PLEKHO2 contribute to diabetic retinopathy (DR) development. Functional prediction analysis strengthened the likelihood of participation of PLEKHO2 in the development of DR (Eric C. Han et al., 2012).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PH domain-containing family O member 2; PH domain-containing family Q member 1; PH domain-containing protein; Pleckstrin homology domain-containing family O member 2; Pleckstrin homology domain-containing family Q member 1; TRIP; unnamed protein product
Gene Aliases: PLEKHO2; PLEKHQ1; PP1628; PP9099
UniProt ID: (Human) Q8TD55
Entrez Gene ID: (Human) 80301
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support