Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide Sequence: SQQIHVGILS PTVDDDDNRC LVDVNSRPRL TECSYAKAKR MKLHWQFSQG GPIQNRKSKR CLELQENSDL EFGFQLVLQK CSGQHWSITN VLRSL*
Catalyzes the initial reaction in O-linked oligosaccharide biosynthesis, the transfer of an N-acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GalNAc-T-like protein 4; GalNAc-T18; GalNAc-transferase 18; Polypeptide GalNAc transferase 18; Polypeptide GalNAc transferase-like protein 4; Polypeptide N-acetylgalactosaminyltransferase 18; Polypeptide N-acetylgalactosaminyltransferase-like protein 4; pp-GaNTase-like protein 4; Protein-UDP acetylgalactosaminyltransferase-like protein 4; putative polypeptide N-acetylgalactosaminyltransferase-like protein 4; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase-like protein 4; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 15; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 18; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase-like 4; unnamed protein product
Gene Aliases: GalNAc-T15; GalNAc-T18; GALNACT18; GALNT15; GALNT18; GALNTL4
UniProt ID: (Human) Q6P9A2
Entrez Gene ID: (Human) 374378
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support