Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ZFP36 Antibodies

          Invitrogen

          ZFP36 Monoclonal Antibody (OTI1A2)

          Advanced Verification
          View all (32) ZFP36 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ZFP36 Monoclonal Antibody (OTI1A2)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (12)
          • Advanced Verification (2)
          ZFP36 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ZFP36 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 14

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ZFP36 Antibody (MA5-25406) in ICC/IF

          Immunofluorescence analysis of ZFP36 was performed using 70% confluent log phase RAW 264.7 cells treated with LPS (10 ng/mL for 2hours). The cells were fixed with 4% Paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 2% BSA for 10 minutes at room temperature. The cells were labeled with ZFP36 Monoclonal Antibody (OTI1A2) (Product # MA5-25406) at 1:100 dilution in 0.1% BSA, incubated at 4 degree celsius overnight and then labeled with Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Seco... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ZFP36 Antibody in Immunocytochemistry (ICC/IF)
          ZFP36 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ZFP36 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ZFP36 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ZFP36 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ZFP36 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ZFP36 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ZFP36 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ZFP36 Antibody in Western Blot (WB)
          ZFP36 Antibody in Western Blot (WB)
          ZFP36 Antibody in Western Blot (WB)
          ZFP36 Antibody in Western Blot (WB)
          ZFP36 Antibody
          ZFP36 Antibody
          ZFP36 Monoclonal Antibody (OTI1A2)

          Product Details

          MA5-25406

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:150
          -

          Immunocytochemistry (ICC/IF)

          1:100
          -

          Flow Cytometry (Flow)

          1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          OTI1A2

          Immunogen

          Full length human recombinant protein of ZFP36 produced in HEK293T cell
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 1% BSA, 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2725702

          Target Information

          ZFP36 is a probable regulatory protein with a novel zinc finger structure involved in regulating the response to growth factors. Knockdown of ZFP36 increased both cognate macrophage gene mRNAs and inflammatory tumor necrosis factor protein release. Overexpression of ZFP36 resulted in decreased levels of the same genes supporting its role in regulating macrophage gene expression. Multiple phosphorylation sites in ZFP36 in mammalian cells were reported. ZFP36 can be phosphorylated by JNK as well as by the other members of the greater MAP kinase family. Gene expression profiling predicts clinical outcome of prostate cancer. Inappropriate ZFP36 production may be one factor that contributes to higher rheumatoid arthritis disease activity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: G0/G1 switch regulatory protein 24; Growth factor-inducible nuclear protein NUP475; mRNA decay activator protein ZFP36; TPA-induced sequence 11; Tristetraprolin; tristetraproline; unnamed protein product; Zfp-36; Zinc finger protein 36; zinc finger protein 36 homolog; zinc finger protein 36, C3H type, homolog

          View more View less

          Gene Aliases: G0S24; GOS24; NUP475; RNF162A; TIS11; TIS11A; TIS11D; TISII; TTP; Zfp-36; ZFP36

          View more View less

          UniProt ID: (Human) P26651, (Mouse) P22893

          View more View less

          Entrez Gene ID: (Human) 7538, (Mouse) 22695

          View more View less

          Function(s)
          DNA binding RNA binding mRNA binding mRNA 3'-UTR binding protein binding zinc ion binding enzyme binding protein kinase binding C-C chemokine binding heat shock protein binding mRNA 3'-UTR AU-rich region binding macromolecular complex binding metal ion binding RNA polymerase binding 14-3-3 protein binding protein-RNA sequence-specific adaptor activity AU-rich element binding poly(A) RNA binding RNA metabolism protein
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter MAPK cascade nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay nuclear-transcribed mRNA poly(A) tail shortening mRNA catabolic process response to wounding regulation of keratinocyte proliferation gene silencing by RNA nuclear-transcribed mRNA catabolic process, deadenylation-independent decay regulation of tumor necrosis factor production negative regulation of interleukin-2 production negative regulation of viral transcription miRNA mediated inhibition of translation p38MAPK cascade response to starvation regulation of mRNA stability cellular response to fibroblast growth factor stimulus positive regulation of fat cell differentiation regulation of keratinocyte differentiation negative regulation of erythrocyte differentiation mRNA transport positive regulation of nuclear-transcribed mRNA poly(A) tail shortening 3'-UTR-mediated mRNA destabilization 3'-UTR-mediated mRNA stabilization cellular response to lipopolysaccharide cellular response to tumor necrosis factor cellular response to epidermal growth factor stimulus cellular response to glucocorticoid stimulus cellular response to granulocyte macrophage colony-stimulating factor stimulus negative regulation of cytokine production involved in inflammatory response positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay positive regulation of deadenylation-independent decapping of nuclear-transcribed mRNA regulation of keratinocyte apoptotic process negative regulation of polynucleotide adenylyltransferase activity positive regulation of intracellular mRNA localization negative regulation of 3'-UTR-mediated mRNA stabilization positive regulation of gene silencing by miRNA regulation of transcription from RNA polymerase II promoter intracellular signal transduction negative regulation of myeloid cell differentiation negative regulation of inflammatory response RNA destabilization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline