Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The protein encoded by this gene is cilium-specific and is required for the stability of the ciliary architecture. It is involved in the regulation of microtubule-based cilia and actin-based brush border microvilli. Mutations in this gene are associated with primary ciliary dyskinesia-13.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dynein assembly factor 1, axonemal; Dynein axonemal assembly factor 1; leucine rich repeat containing 50; Leucine-rich repeat-containing protein 50; outer row dynein assembly 7 homolog; testicular tissue protein Li 110; unnamed protein product
Gene Aliases: 4930457P18Rik; CILD13; DAU1; DNAAF1; LRRC50; ODA7; RGD1310542; swt
UniProt ID: (Human) Q8NEP3, (Mouse) Q9D2H9, (Rat) Q6AYH9
Entrez Gene ID: (Human) 123872, (Mouse) 68270, (Rat) 361419
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support