Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The gene CCDC27 is part of a family of genes that encode coiled-coil domain-containing proteins. These proteins are characterized by their structural motif-a coiled-coil domain, which consists of two or more alpha helices coiled together. While specific details about CCDC27 are not thoroughly documented in the literature, coiled-coil domain proteins are known for their diverse roles in cellular structure and function, including participation in intracellular transport, signal transduction, and cellular scaffolding. Although the role of CCDC27 within these processes remains speculative, similar coiled-coil domain proteins are implicated in cancer pathogenesis due to their involvement in cellular processes that are frequently dysregulated in oncogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coiled-coil domain-containing protein 27; RP1-286D6.1; unnamed protein product
Gene Aliases: CCDC27
UniProt ID: (Human) Q2M243
Entrez Gene ID: (Human) 148870
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support