Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • BMP4 Antibodies

          Invitrogen

          BMP-4 Monoclonal Antibody (02)

          View all (31) BMP4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite BMP-4 Monoclonal Antibody (02)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          BMP-4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          BMP-4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          BMP-4 Antibody (MA5-29056) in ICC/IF

          Immunofluorescence staining of Human BMP-4 in Hela cells. Cells were fixed with 4% PFA, permeabilzed with 1% Triton X-100 in PBS, blocked with 10% serum, and incubated with BMP-4 Monoclonal Antibody (2) (Product # MA5-29056, 1:60) at 37°C 1 hour. Then cells were stained with the Alexa Fluor® 488-conjugated Goat Anti-mouse IgG secondary antibody (green) and counterstained with DAPI (blue). Positive staining was localized to cytoplasm. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          BMP-4 Antibody in Immunocytochemistry (ICC/IF)
          BMP-4 Antibody in Flow Cytometry (Flow)

          Product Details

          MA5-29056

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1:20-1:100
          -

          Flow Cytometry (Flow)

          1:25-1:100
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgM

          Class

          Monoclonal

          Type

          Antibody

          Clone

          02

          Immunogen

          Recombinant Human BMP-4 protein

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2784989

          Product Specific Information

          This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).

          This antibody has specificity for Human BMP-4.

          Target Information

          The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5' untranslated region of this gene has been described and three variants are described, all encoding an identical protein.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: BMP; BMP-2B; BMP-4; Bone morphogenetic protein; Bone morphogenetic protein 2B; Bone morphogenetic protein 4; unnamed protein product

          View more View less

          Gene Aliases: BMP2B; BMP2B1; BMP4; DVR4; MCOPS6; OFC11; ZYME

          View more View less

          UniProt ID: (Human) P12644

          View more View less

          Entrez Gene ID: (Human) 652

          View more View less

          Function(s)
          cytokine activity protein binding growth factor activity heparin binding co-receptor binding chemoattractant activity protein serine/threonine kinase activator activity BMP receptor binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter skeletal system development ossification angiogenesis blood vessel development osteoblast differentiation metanephros development ureteric bud development branching involved in ureteric bud morphogenesis mesoderm formation organ induction kidney development mesonephros development neural tube closure positive regulation of protein phosphorylation positive regulation of endothelial cell proliferation vasculature development endochondral ossification blood vessel endothelial cell proliferation involved in sprouting angiogenesis chondrocyte differentiation hematopoietic progenitor cell differentiation lymphoid progenitor cell differentiation myeloid leukocyte differentiation heart morphogenesis renal system process heart induction secondary heart field specification outflow tract septum morphogenesis membranous septum morphogenesis muscular septum morphogenesis outflow tract morphogenesis sinoatrial node development aortic valve morphogenesis pulmonary valve morphogenesis endocardial cushion development epithelial to mesenchymal transition involved in endocardial cushion formation cardiac right ventricle morphogenesis apoptotic process involved in endocardial cushion morphogenesis cardiac septum development type B pancreatic cell development transcription from RNA polymerase II promoter germ cell development endoderm development mesoderm development mesodermal cell fate determination heart development cell proliferation positive regulation of cell proliferation negative regulation of cell proliferation post-embryonic development anterior/posterior axis specification specification of organ position regulation of cell fate commitment gene expression regulation of gene expression positive regulation of gene expression negative regulation of gene expression positive regulation of epithelial to mesenchymal transition telencephalon development dorsal/ventral neural tube patterning telencephalon regionalization pituitary gland development negative regulation of cell-cell adhesion cell differentiation erythrocyte differentiation monocyte differentiation macrophage differentiation lung development embryonic limb morphogenesis positive regulation of cell migration positive regulation of bone mineralization BMP signaling pathway regulation of BMP signaling pathway positive regulation of BMP signaling pathway negative regulation of BMP signaling pathway positive regulation of epithelial cell differentiation forebrain development positive regulation of protein binding negative regulation of chondrocyte differentiation positive regulation of collagen biosynthetic process negative regulation of T cell differentiation in thymus negative regulation of immature T cell proliferation in thymus embryonic hindlimb morphogenesis tendon cell differentiation deltoid tuberosity development ameloblast differentiation regulation of protein import into nucleus odontogenesis of dentin-containing tooth odontogenesis regulation of odontogenesis of dentin-containing tooth auditory receptor cell differentiation embryonic digit morphogenesis camera-type eye development positive regulation of apoptotic process positive regulation of programmed cell death cell fate commitment regulation of cell differentiation negative regulation of cell differentiation positive regulation of endothelial cell differentiation positive regulation of epidermal cell differentiation negative regulation of myoblast differentiation positive regulation of neuron differentiation positive regulation of osteoblast differentiation positive regulation of ossification negative regulation of cell cycle negative regulation of mitotic nuclear division negative regulation of striated muscle tissue development negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter lung alveolus development mesodermal cell differentiation intermediate mesodermal cell differentiation camera-type eye morphogenesis embryonic morphogenesis anatomical structure formation involved in morphogenesis regulation of smooth muscle cell proliferation positive regulation of smooth muscle cell proliferation negative regulation of smooth muscle cell proliferation neuron fate commitment embryonic cranial skeleton morphogenesis embryonic skeletal system morphogenesis embryonic skeletal system development smooth muscle tissue development stem cell differentiation epithelial cell proliferation positive regulation of epithelial cell proliferation negative regulation of epithelial cell proliferation positive chemotaxis smooth muscle cell differentiation regulation of smooth muscle cell differentiation cartilage development negative regulation of multicellular organismal process cardiac muscle cell differentiation positive regulation of cardiac muscle fiber development lens induction in camera-type eye embryonic skeletal joint morphogenesis bone development cranial suture morphogenesis positive regulation of SMAD protein import into nucleus lung morphogenesis lung vasculature development epithelium development bronchus development trachea development trachea formation epithelial tube branching involved in lung morphogenesis branching involved in prostate gland morphogenesis bud elongation involved in lung branching epithelial cell proliferation involved in lung morphogenesis bud dilation involved in lung branching prostate gland morphogenesis prostatic bud formation muscle tissue development mammary gland formation epithelial-mesenchymal cell signaling negative regulation of prostatic bud formation regulation of branching involved in prostate gland morphogenesis regulation of morphogenesis of a branching structure coronary vasculature development regulation of cartilage development positive regulation of cartilage development positive regulation of branching involved in lung morphogenesis pharyngeal arch artery morphogenesis negative regulation of thymocyte apoptotic process positive regulation of ERK1 and ERK2 cascade cellular response to growth factor stimulus cellular response to BMP stimulus glomerular capillary formation negative regulation of glomerular mesangial cell proliferation nephric duct formation ureter morphogenesis negative regulation of mesenchymal cell proliferation involved in ureter development regulation of branching involved in ureteric bud morphogenesis negative regulation of branching involved in ureteric bud morphogenesis negative regulation of glomerulus development positive regulation of protein localization to nucleus positive regulation of p38MAPK cascade positive regulation of odontoblast differentiation regulation of miRNA transcription negative regulation of pri-miRNA transcription from RNA polymerase II promoter pericyte cell differentiation negative regulation of vascular smooth muscle cell proliferation negative regulation of vascular associated smooth muscle cell migration cardiac jelly development positive regulation of cardiac neural crest cell migration involved in outflow tract morphogenesis regulation of mesodermal cell differentiation negative regulation of metanephric S-shaped body morphogenesis negative regulation of metanephric comma-shaped body morphogenesis positive regulation of primary miRNA processing regulation of cardiac muscle cell differentiation negative regulation of extrinsic apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline