Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 10
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
PSO4 is the human homolog of yeast Pso4, a gene essential for cell survival and DNA repair.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPso4; Neuronal differentiation-related gene protein; Nuclear matrix protein 200; nuclear matrix protein NMP200 related to splicing factor PRP19; nuclear matrix protein SNEV; Pre-mRNA-processing factor 19; prp19 beta; PRP19/PSO4 homolog; PRP19/PSO4 pre-mRNA processing factor 19 homolog; psoralen 4; RING-type E3 ubiquitin transferase PRP19; Senescence evasion factor
Gene Aliases: AA617263; AL024362; D19Wsu55e; hPSO4; NMP200; PRP19; PRPF19; PSO4; SNEV; UBOX4
UniProt ID: (Human) Q9UMS4, (Mouse) Q99KP6, (Rat) Q9JMJ4
Entrez Gene ID: (Human) 27339, (Mouse) 28000, (Rat) 246216
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support