Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
PGC-1 beta is a tissue specific co-activator that enhances the activity of estrogen receptor alpha. The co-activation function relies on a bipartite transcriptional activation domain and two LXXLL motifs that interact with the AR2 domain of ER-alpha in an estrogen-dependent manner. Ectopic expression of this protein has been shown to lead to increased mitochondrial number and basal oxygen consumption.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ERR ligand 1; L-PGC-1alpha; Ligand effect modulator 6; ligand effect modulator-6; PERC; PERC-s; lacks one estrogen receptor interaction domain; peroxisome proliferative activated receptor gamma coactivator 1; peroxisome proliferative activated receptor, gamma, coactivator 1 alpha; peroxisome proliferative activated receptor, gamma, coactivator 1 beta; peroxisome proliferator-activated receptor gamma coactivator 1 beta; Peroxisome proliferator-activated receptor gamma coactivator 1-alpha; Peroxisome proliferator-activated receptor gamma coactivator 1-beta; peroxisome proliferator-activated receptor gamma coactivator 1beta-2a; peroxisome proliferator-activated receptor gamma coactivator-1 alpha; peroxisome proliferator-activated receptor gamma, coactivator 1 beta; pgc-1 alpha; pgc-1 beta; PGC-1-alpha; PGC-1-beta; PGC-1-related estrogen receptor alpha coactivator; PGC-1alpha; Pgc-1alphaa; PGC-1beta; PGC-1beta/ERRL1; PGC1beta-1a; PGC1beta-1b; PGC1beta-2a; PGC1beta-2b; PPAR gamma coactivator variant form; PPAR Gamma Coactivator-1; PPAR gamma coactivator-1beta protein; PPAR-gamma coactivator 1-alpha; PPAR-gamma coactivator 1-beta; PPARAGCIalpha; PPARAGCIbeta; PPARgamma coactivator 1 beta; PPARgamma coactivator 1alpha; PPARGC-1-alpha; PPARGC-1-beta; transcriptional coactivator; transcriptional coactivator; PGC-1; unnamed protein product
Gene Aliases: 4631412G21Rik; A830037N07Rik; ENSMUSG00000079510; ERRL1; Gm11133; LEM6; PERC; PGC-1; PGC-1(alpha); PGC-1(beta); PGC-1alpha; PGC-1v; PGC1; PGC1A; PGC1B; PGC1beta; Pgco1; PPARGC1; PPARGC1A; PPARGC1B
UniProt ID: (Human) Q9UBK2, (Human) Q86YN6, (Mouse) O70343, (Rat) Q9QYK2, (Rat) Q811R2, (Mouse) Q8VHJ7
Entrez Gene ID: (Human) 10891, (Human) 133522, (Mouse) 19017, (Rat) 83516, (Rat) 291567, (Mouse) 170826
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support