Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
MA5-36218 reacts to human TRK. This antibody may also react to mouse or rat TRK, as predicted by immunogen homology.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BDNF-tropomyosine receptor kinase B; BDNF/NT-3 growth factors receptor; ETS related protein-neurotrophic receptor tyrosine kinase fusion protein; ETV6-NTRK3 fusion; gp140trk; GP145-TrkB; GP145-TrkC; High affinity nerve growth factor receptor; Neurotrophic tyrosine kinase receptor type 1; Neurotrophic tyrosine kinase receptor type 2; Neurotrophic tyrosine kinase receptor type 3; neurotrophic tyrosine kinase, receptor, type 1; neurotrophic tyrosine kinase, receptor, type 3; NT-3 growth factor receptor; NTRK2; NTRK3; Oncogene TRK; p140-TrkA; splice variant; TEL-TrkC fusion protein; trk tyrosine-specific protein kinase; Trk-A; Trk-B; Trk-C; TRK1-transforming tyrosine kinase protein; TrkB; TrkB tyrosine kinase; TrkB-T-Shc; trkC; TrkC tyrosine kinase; tropomyosin receptor kinase A; Tropomyosin-related kinase A; Tropomyosin-related kinase B; truncated trkB; Tyrosine kinase receptor; Tyrosine kinase receptor A; tyrosine kinase receptor B; tyrosine kinase receptor C; unnamed protein product
Gene Aliases: DEE58; EIEE58; gp145(trkC); GP145-TrkB; GP145-TrkC; MTC; NTRK1; NTRK2; NTRK3; OBHD; p140-TrkA; TRK; Trk-A; trk-B; TRK1; TRKA; TRKB; TRKC
UniProt ID: (Human) P04629, (Human) Q16620, (Human) Q16288
Entrez Gene ID: (Human) 4914, (Human) 4915, (Human) 4916
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support