Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PI3K p85/p55 Antibodies

          Invitrogen

          Phospho-PI3K p85/p55 (Tyr458, Tyr199) Recombinant Rabbit Monoclonal Antibody (PI3KY458-1A11), PE

          View all (13) PI3K p85/p55 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Phospho-PI3K p85/p55 (Tyr458, Tyr199) Recombinant Rabbit Monoclonal Antibody (PI3KY458-1A11), PE

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Phospho-PI3K p85/p55 (Tyr458, Tyr199) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          Phospho-PI3K p85/p55 (Tyr458, Tyr199) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-PI3K p85/p55 (Tyr458, Tyr199) Antibody (MA5-36954) in Flow

          Flow cytometry of Phospho-PI3K p85/p55 (Tyr458, Tyr199) using Ramos cells. Samples were incubated in monoclonal Phospho-PI3K p85/p55 (Tyr458, Tyr199) antibody (Product # MA5-36954). Images correspond with the following conditions: unstained (blue), untreated (red), treated with pervanadate (green). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-PI3K p85/p55 (Tyr458, Tyr199) Antibody in Flow Cytometry (Flow)
          Phospho-PI3K p85/p55 (Tyr458, Tyr199) Recombinant Rabbit Monoclonal Antibody (PI3KY458-1A11), PE

          Product Details

          MA5-36954

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          5 µL/1x10^6 cells
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG, kappa

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          PI3KY458-1A11

          Immunogen

          A synthetic phospho-peptide corresponding to residues surrounding Tyr458 of human phospho PI3K p85.
          3D Epitope / Immunogen

          Conjugate

          PE PE PE
          • Unconjugated
          • APC 
          • FITC
          • Request custom conjugation

          Excitation/Emission Max

          565/576 nm View spectra spectra

          Form

          Liquid

          Purification

          Protein A/G

          Storage buffer

          PBS, pH 7.4, with 0.2% BSA

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2896889

          Product Specific Information

          Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

          Target Information

          Phosphoinositide 3-kinase (PI3K, phospho PI3K) Binds to activated (phosphorylated) protein-Tyr kinases, through its SH2 domain, and acts as an adapter, mediating the association of the p110 catalytic unit to the plasma membrane. Phospho PI3K is necessary for the insulin-stimulated increase in glucose uptake and glycogen synthesis in insulin-sensitive tissues. During insulin stimulation, it also binds to IRS-1. Growth factors and hormones trigger this phosphorylation event, which in turn coordinates cell growth, cell cycle entry, cell migration, and cell survival.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: growth factor receptor bound 1; OTTHUMP00000009783; OTTHUMP00000009784; p55PIK; p85-alpha; GRB1; Phosphatidylinositol 3-kinase 55 kDa regulatory subunit gamma; Phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha; Phosphatidylinositol 3-kinase 85 kDa regulatory subunit beta; Phosphatidylinositol 3-kinase regulatory subunit alpha; Phosphatidylinositol 3-kinase regulatory subunit beta; Phosphatidylinositol 3-kinase regulatory subunit gamma; phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha); phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 2 (p85 beta); phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 3 (p55, gamma); phosphatidylinositol 3-kinase-associated p-85 alpha; phosphoinositide-3-kinase regulatory subunit alpha; phosphoinositide-3-kinase regulatory subunit beta; phosphoinositide-3-kinase, regulatory subunit 1 (alpha); phosphoinositide-3-kinase, regulatory subunit 2 (beta); phosphoinositide-3-kinase, regulatory subunit 3 (gamma); PI3 kinase-associated p85; PI3-kinase regulatory subunit alpha; PI3-kinase regulatory subunit beta; PI3-kinase regulatory subunit gamma; PI3-kinase subunit p55-gamma; PI3-kinase subunit p85-alpha; PI3-kinase subunit p85-beta; PI3K regulatory subunit alpha; PI3K regulatory subunit beta; PI3K regulatory subunit gamma; PI3K-p46; polypeptide 3; ptdIns-3-kinase regulatory subunit alpha; ptdIns-3-kinase regulatory subunit beta; ptdIns-3-kinase regulatory subunit gamma; ptdIns-3-kinase regulatory subunit p55-gamma; PtdIns-3-kinase regulatory subunit p85-al; ptdIns-3-kinase regulatory subunit p85-beta; RP4-533D7.2; unnamed protein product

          View more View less

          Gene Aliases: AA414954; AGM7; GRB1; IMD36; MPPH; MPPH1; p50alpha; p55; p55-GAMMA; p55alpha; p55PIK; p85; p85-ALPHA; p85-BETA; p85alpha; P85B; p85beta; PI3K; PIK3R1; PIK3R2; PIK3R3

          View more View less

          UniProt ID: (Human) P27986, (Human) O00459, (Human) Q92569, (Mouse) P26450, (Mouse) O08908, (Mouse) Q64143

          View more View less

          Entrez Gene ID: (Human) 5295, (Human) 5296, (Human) 8503, (Mouse) 18708, (Mouse) 18709, (Mouse) 18710

          View more View less

          Function(s)
          phosphotyrosine binding transmembrane receptor protein tyrosine kinase adaptor activity GTPase activator activity insulin receptor binding insulin-like growth factor receptor binding neurotrophin TRKA receptor binding protein binding kinase regulator activity kinase activator activity protein phosphatase binding phosphatidylinositol 3-kinase regulator activity phosphatidylinositol 3-kinase regulatory subunit binding ErbB-3 class receptor binding phosphatidylinositol 3-kinase binding insulin binding insulin receptor substrate binding 1-phosphatidylinositol-3-kinase regulator activity protein heterodimerization activity phosphatidylinositol kinase activity enzyme-substrate adaptor activity phosphatidylinositol 3-kinase activator activity receptor tyrosine kinase binding 1-phosphatidylinositol-3-kinase activity receptor binding platelet-derived growth factor receptor binding calmodulin binding protein C-terminus binding transcription factor binding protein kinase binding protein domain specific binding estrogen receptor binding ubiquitin protein ligase binding ATPase binding phosphoprotein binding kinase modulator
          Process(es)
          cellular glucose homeostasis negative regulation of cell-matrix adhesion positive regulation of leukocyte migration transcription from RNA polymerase II promoter protein import into nucleus immune response negative regulation of cell adhesion signal transduction insulin receptor signaling pathway extrinsic apoptotic signaling pathway via death domain receptors intrinsic apoptotic signaling pathway in response to DNA damage positive regulation of lamellipodium assembly protein transport cytokine-mediated signaling pathway B cell differentiation T cell differentiation osteoclast differentiation positive regulation of tumor necrosis factor production cellular response to insulin stimulus positive regulation of RNA splicing regulation of toll-like receptor 4 signaling pathway substrate adhesion-dependent cell spreading cellular response to UV response to endoplasmic reticulum stress interleukin-18-mediated signaling pathway natural killer cell mediated cytotoxicity positive regulation of protein import into nucleus negative regulation of apoptotic process protein kinase B signaling negative regulation of osteoclast differentiation positive regulation of transcription from RNA polymerase II promoter positive regulation of glucose import phosphatidylinositol phosphorylation insulin-like growth factor receptor signaling pathway positive regulation of smooth muscle cell proliferation protein stabilization positive regulation of filopodium assembly regulation of stress fiber assembly negative regulation of stress fiber assembly growth hormone receptor signaling pathway T follicular helper cell differentiation myeloid leukocyte migration positive regulation of focal adhesion disassembly positive regulation of endoplasmic reticulum unfolded protein response regulation of protein localization to plasma membrane positive regulation of protein localization to plasma membrane regulation of autophagy regulation of actin filament polymerization negative regulation of MAPK cascade positive regulation of cell adhesion cell migration involved in sprouting angiogenesis positive regulation of gene expression positive regulation of cell migration phosphatidylinositol-3-phosphate biosynthetic process negative regulation of anoikis positive regulation of protein phosphorylation glucose metabolic process protein phosphorylation transport negative regulation of heart rate negative regulation of muscle cell apoptotic process phosphatidylinositol 3-kinase signaling negative regulation of cell-cell adhesion response to insulin positive regulation of transcription factor import into nucleus regulation of phosphatidylinositol 3-kinase activity positive regulation of myoblast differentiation negative regulation of blood pressure negative regulation of proteolysis negative regulation of smooth muscle cell proliferation response to glucocorticoid NFAT protein import into nucleus response to cAMP regulation of establishment of protein localization to plasma membrane regulation of phosphorylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline