Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Forms hydroxylysine residues in -Xaa-Lys-Gly- sequences in collagens. These hydroxylysines serve as sites of attachment for carbohydrate units and are essential for the stability of the intermolecular collagen cross-links.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LH1; lysine hydroxylase; lysyl hydroxlase 1; Lysyl hydroxylase 1; procollagen lysyl hydroxylase 1; procollagen-lysine 1, 2-oxoglutarate 5-dioxygenase 1; Procollagen-lysine,2-oxoglutarate 5-dioxygenase 1; unnamed protein product
Gene Aliases: EDS6; EDSKCL1; LH; LH1; LLH; PLOD; PLOD1
UniProt ID: (Human) Q02809
Entrez Gene ID: (Human) 5351
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support