Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive Samples: 293T, A-549, SH-SY5Y, Mouse brain, Rat brain
Immunogen sequence: KEMIRSRKAV SKLYASKAHM NSVLMGMKNQ LAVLRVAGSL QKSTEVMKAM QSLVKIPEIQ ATMRELSKEM MKAGIIEEML EDTFESMDDQ EEMEEEAEME IDRILFEITA GALGKAPSKV TDALPEPEPP GAMAASEDEE EEEEALEAMQ SRLATL
Probable core component of the endosomal sorting required for transport complex III (ESCRT-III) which is involved in multivesicular bodies (MVBs) formation and sorting of endosomal cargo proteins into MVBs. MVBs contain intraluminal vesicles (ILVs) that are generated by invagination and scission from the limiting membrane of the endosome and mostly are delivered to lysosomes enabling degradation of membrane proteins, such as stimulated growth factor receptors, lysosomal enzymes and lipids. The MVB pathway appears to require the sequential function of ESCRT-O, -I,-II and -III complexes. ESCRT-III proteins mostly dissociate from the invaginating membrane before the ILV is released. The ESCRT machinery also functions in topologically equivalent membrane fission events, such as the terminal stages of cytokinesis and the budding of enveloped viruses (HIV-1 and other lentiviruses). ESCRT-III proteins are believed to mediate the necessary vesicle extrusion and/or membrane fission activities, possibly in conjunction with the AAA ATPase VPS4. Selectively binds to phosphatidylinositol 3,5-bisphosphate PtdIns(3,5)P2 and PtdIns(3,4)P2 in preference to other phosphoinositides tested. Involved in late stages of cytokinesis. Plays a role in endosomal sorting/trafficking of EGF receptor. Isoform 2 prevents stress- mediated cell death and accumulation of reactive oxygen species when expressed in yeast cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 25.1 protein; CGI-149; Charged multivesicular body protein 3; CHMP family, member 3; Chromatin-modifying protein 3; hVps24; NEDF; Neuroendocrine differentiation factor; rVps24p; unnamed protein product; vacuolar protein sorting 24; vacuolar protein sorting 24 homolog; Vacuolar protein sorting-associated protein 24; vacuolar protein-sorting-associated protein 24; Vps24p protein
Gene Aliases: 25.1; 4921505F14Rik; 9130011K15Rik; CGI-149; CGI-49; CGI149; CHMP3; D6Ertd286e; NEDF; VPS24
UniProt ID: (Human) Q9Y3E7, (Rat) Q8CGS4, (Mouse) Q9CQ10
Entrez Gene ID: (Human) 51652, (Rat) 282834, (Mouse) 66700
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support