Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 5
Positive Samples: HeLa, K-562, SH-SY5Y, Mouse testis
Immunogen sequence: KTLTDEDEAD QGGWLAAFWK DFRRRFLALL SYQFSTFSPS LALNIIQNRN MGKPAQPALS REELEALFLP YDLKRLEMYS RNMVDYHLIM DMIPAISRIY FLNQLGDLAL SAAQSALLLG IGLQHKSVDQ LEKEIELPSG QLMGLFNRII RKVVKLFNEV QEKAIEEQMV AAKDVVMEPT MKTLSDDLDE AAKEFQEKHK KEVGKLKSMD LSEYIIRGDD EEWNEVLNKA GPNASIISLK SDKKRKLEAK QEPKQSKKLK NRETKNKKDM KLKRKK
IRS4 encodes the insulin receptor substrate 4, a cytoplasmic protein that contains many potential tyrosine and serine/threonine phosphorylation sites. Tyrosine-phosphorylated IRS4 protein has been shown to associate with cytoplasmic signalling molecules that contain SH2 domains. The IRS4 protein is phosphorylated by the insulin receptor tyrosine kinase upon receptor stimulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 18S rRNA cytosine acetyltransferase; hALP; N-acetyltransferase 10; N-acetyltransferase 10 (GCN5-related); N-acetyltransferase-like protein; novel putative ATPase domain containing protein; RNA cytidine acetyltransferase; unnamed protein product
Gene Aliases: AI429152; ALP; KIAA1709; Kre33; NAT10; NET43; RGD1306717
UniProt ID: (Human) Q9H0A0, (Mouse) Q8K224
Entrez Gene ID: (Human) 55226, (Mouse) 98956, (Rat) 311257
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support