Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • PGC1 alpha/beta Antibodies

          Invitrogen

          PGC1 alpha/beta Recombinant Rabbit Monoclonal Antibody (5K0F4)

          View all (4) PGC1 alpha/beta antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PGC1 alpha/beta Recombinant Rabbit Monoclonal Antibody (5K0F4)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          PGC1 alpha/beta Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PGC1 alpha/beta Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PGC1 alpha/beta Antibody (MA5-37948) in WB

          Western blot analysis of PGC1 alpha/beta in extracts of various cell lines. Samples were incubated with PGC1 alpha/beta Monoclonal antibody (Product # MA5-37948) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Basic Kit. Exposure time: 1s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PGC1 alpha/beta Antibody in Western Blot (WB)
          PGC1 alpha/beta Recombinant Rabbit Monoclonal Antibody (5K0F4)

          Product Details

          MA5-37948

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:6,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          5K0F4

          Immunogen

          Synthetic peptide
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          2 mg/mL

          Amount

          200 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 0.05% BSA, 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2897868

          Product Specific Information

          Positive Samples: Mouse liver, Rat heart

          Immunogen sequence: VFGEIEECTV NLRDDGDSYG FITYRYTCDA FAALENGYTL RRSNETDFEL YFCGRKQFFK SNYADLDSNS DDFDPASTKS KYDSLDFDSL LKEAQRSLRR

          Target Information

          PGC-1 beta is a tissue specific co-activator that enhances the activity of estrogen receptor alpha. The co-activation function relies on a bipartite transcriptional activation domain and two LXXLL motifs that interact with the AR2 domain of ER-alpha in an estrogen-dependent manner. Ectopic expression of this protein has been shown to lead to increased mitochondrial number and basal oxygen consumption.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ERR ligand 1; L-PGC-1alpha; Ligand effect modulator 6; ligand effect modulator-6; PERC; PERC-s; lacks one estrogen receptor interaction domain; peroxisome proliferative activated receptor gamma coactivator 1; peroxisome proliferative activated receptor, gamma, coactivator 1 alpha; peroxisome proliferative activated receptor, gamma, coactivator 1 beta; peroxisome proliferator-activated receptor gamma coactivator 1 beta; Peroxisome proliferator-activated receptor gamma coactivator 1-alpha; Peroxisome proliferator-activated receptor gamma coactivator 1-beta; peroxisome proliferator-activated receptor gamma coactivator 1beta-2a; peroxisome proliferator-activated receptor gamma coactivator-1 alpha; peroxisome proliferator-activated receptor gamma, coactivator 1 beta; pgc-1 alpha; pgc-1 beta; PGC-1-alpha; PGC-1-beta; PGC-1-related estrogen receptor alpha coactivator; PGC-1alpha; Pgc-1alphaa; PGC-1beta; PGC-1beta/ERRL1; PGC1beta-1a; PGC1beta-1b; PGC1beta-2a; PGC1beta-2b; PPAR gamma coactivator variant form; PPAR Gamma Coactivator-1; PPAR gamma coactivator-1beta protein; PPAR-gamma coactivator 1-alpha; PPAR-gamma coactivator 1-beta; PPARAGCIalpha; PPARAGCIbeta; PPARgamma coactivator 1 beta; PPARgamma coactivator 1alpha; PPARGC-1-alpha; PPARGC-1-beta; transcriptional coactivator; transcriptional coactivator; PGC-1; unnamed protein product

          View more View less

          Gene Aliases: 4631412G21Rik; A830037N07Rik; ENSMUSG00000079510; ERRL1; Gm11133; LEM6; PERC; PGC-1; PGC-1(alpha); PGC-1(beta); PGC-1alpha; PGC-1v; PGC1; PGC1A; PGC1B; PGC1beta; Pgco1; PPARGC1; PPARGC1A; PPARGC1B

          View more View less

          UniProt ID: (Human) Q9UBK2, (Human) Q86YN6, (Mouse) O70343, (Rat) Q9QYK2, (Rat) Q811R2, (Mouse) Q8VHJ7

          View more View less

          Entrez Gene ID: (Human) 10891, (Human) 133522, (Mouse) 19017, (Rat) 83516, (Rat) 291567, (Mouse) 170826

          View more View less

          Function(s)
          nucleic acid binding DNA binding transcription cofactor activity transcription coactivator activity RNA binding protein binding ligand-dependent nuclear receptor binding chromatin DNA binding ubiquitin protein ligase binding sequence-specific DNA binding RNA polymerase II sequence-specific DNA binding transcription factor binding lncRNA binding DNA-binding transcription factor binding estrogen receptor binding AF-2 domain binding nucleotide binding RNA polymerase II transcription cofactor activity chromatin binding transcription factor binding ligand-dependent nuclear receptor transcription coactivator activity molecular_function receptor activator activity transcription cofactor
          Process(es)
          temperature homeostasis cellular glucose homeostasis response to dietary excess gluconeogenesis regulation of transcription, DNA-templated transcription initiation from RNA polymerase II promoter mRNA processing mitochondrion organization digestion RNA splicing positive regulation of gene expression response to muscle activity fatty acid oxidation respiratory electron transport chain developmental process circadian regulation of gene expression cellular response to oxidative stress homeostatic process response to starvation regulation of circadian rhythm negative regulation of neuron apoptotic process cellular respiration positive regulation of gluconeogenesis positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of fatty acid oxidation rhythmic process negative regulation of smooth muscle cell proliferation protein stabilization brown fat cell differentiation neuron apoptotic process adipose tissue development protein-containing complex assembly energy homeostasis positive regulation of cold-induced thermogenesis intracellular estrogen receptor signaling pathway positive regulation of osteoclast differentiation transcription, DNA-templated regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter positive regulation of mitochondrion organization negative regulation of smooth muscle cell migration positive regulation of cellular metabolic process negative regulation of glycolytic process positive regulation of smooth muscle cell proliferation positive regulation of sequence-specific DNA binding transcription factor activity negative regulation of neuron death positive regulation of cellular respiration positive regulation of mitochondrial DNA metabolic process positive regulation of muscle tissue development negative regulation of receptor activity regulation of N-methyl-D-aspartate selective glutamate receptor activity positive regulation of energy homeostasis positive regulation of ATP biosynthetic process response to hypoxia galactose metabolic process androgen metabolic process response to cold response to organic cyclic compound response to activity response to nutrient levels response to statin response to drug response to leucine flavone metabolic process cellular response to nitrite cellular response to tumor necrosis factor cellular response to fatty acid cellular response to hypoxia response to epinephrine response to norepinephrine cellular response to thyroid hormone stimulus ossification transcription from mitochondrial promoter actin filament organization positive regulation of alkaline phosphatase activity cellular response to reactive oxygen species positive regulation of phosphorylation positive regulation of bone resorption negative regulation of transcription, DNA-templated response to glucocorticoid response to cAMP bone trabecula formation positive regulation of receptor activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline