Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 6
Positive Samples: NIH/3T3, Mouse brain, Mouse thymus
Immunogen sequence: MDPFLVLLHS LSGSLSGNDL MELKFLCRER VSKRKLERVQ SGLDLFTVLL EQNDLERGHT GLLRELLASL RRHDLLQRLD DFEAGTATAA PPGEADLQVA FDIVCDNVGR DWKRLARELK VSEAKMDGIE EKYPRSLSER VRESLKVWKN AEKKNASVAG LVKALRTCRL NLVADLVEEA QESVSKSENM SPVLRDSTVS SSETP
The protein encoded by this gene is an adaptor molecule that interacts with various cell surface receptors and mediates cell apoptotic signals. Through its C-terminal death domain, this protein can be recruited by TNFRSF6/Fas-receptor, tumor necrosis factor receptor, TNFRSF25, and TNFSF10/TRAIL-receptor, and thus it participates in the death signaling initiated by these receptors. Interaction of this protein with the receptors unmasks the N-terminal effector domain of this protein, which allows it to recruit caspase-8, and thereby activate the cysteine protease cascade. Knockout studies in mice also suggest the importance of this protein in early T cell development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FADD; Fas (TNFRSF6)-associated via death domain; FAS-associated death domain protein; FAS-associating death domain-containing protein; Fas-associating protein with death domain; GIG3; Growth-inhibiting gene 3 protein; Mediator of receptor induced toxicity; mediator of receptor-induced toxicity; MGC8528; protein FADD; unnamed protein product
Gene Aliases: FADD; GIG3; IMD90; MORT1; Mort1/FADD
UniProt ID: (Human) Q13158, (Mouse) Q61160
Entrez Gene ID: (Human) 8772, (Mouse) 14082, (Rat) 266610
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support