Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Positive Samples: Raji, SH-SY5Y, Mouse lung, Mouse brain, Rat brain
Immunogen sequence: KGALKQNKEV LELAFSILYD SNCQLNFIAP DKHEYCIWTD GLNALLGKDM MSDLTRNDLD TLLSMEIKLR LLDLENIQIP DAPPPIPKEP SNYDFVYDCN
The protein encoded by this gene interacts with the dedicator of cyto-kinesis 1 protein to promote phagocytosis and effect cell shape changes. Similarity to a C. elegans protein suggests that this protein may function in apoptosis and in cell migration. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ced-12 homolog 1; engulfment and cell motility 1 splice 1; engulfment and cell motility 1, ced-12 homolog; Engulfment and cell motility protein 1; KIAA0281; MGC126406; OTTHUMP00000207056; OTTHUMP00000207060; Protein ced-12 homolog; unnamed protein product
Gene Aliases: 6330578D22Rik; C230095H21Rik; CED-12; CED12; ELMO-1; ELMO1; ELMO1s; KIAA0281
UniProt ID: (Human) Q92556, (Mouse) Q8BPU7
Entrez Gene ID: (Human) 9844, (Mouse) 140580, (Rat) 361251
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support