Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive Samples: SH-SY5Y, Mouse brain
Immunogen sequence: METDAPQPGL ASPDSPHDPC KMFIGGLSWQ TTQEGLREYF GQFGEVKECL VMRDPLTKRS RGFGFVTFMD QAGVDKVLAQ SRHELDSKTI DPKVAFPRRA
Musashi-1 belongs to the Musashi family of RNA-binding proteins which play a role in cell fate determination, and maintenance of stem-cell state. Musashi family proteins are also involved in post transcriptional gene regulation. The gene encodes a protein with two conserved tandem RNA recognition motifs with highly conserved RNP (ribonucleoprotein) motifs. In mammals, Musashi-1 is expressed in fetal kidney, brain, liver and lung, and in adult brain and pancreas. In humans, the gene is located on chromosome 12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Musashi homolog 1(Drosophila); Musashi-1; musashi1; RNA-binding protein Musashi homolog 1; unnamed protein product
Gene Aliases: m-Msi-1; MSI1; Msi1h; Musahi1
UniProt ID: (Human) O43347, (Mouse) Q61474
Entrez Gene ID: (Human) 4440, (Mouse) 17690
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support