Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • DRD1 Antibodies

          Invitrogen

          DRD1 Recombinant Rabbit Monoclonal Antibody (JM10-93)

          View all (37) DRD1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite DRD1 Recombinant Rabbit Monoclonal Antibody (JM10-93)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          DRD1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          DRD1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DRD1 Antibody (MA5-42467) in ICC/IF

          Immunocytochemistry-Immunofluorescence analysis of of DRD1 in SH-SY5Y cells (red). Formalin fixed cells were permeabilized with 0.1% Triton X-100 in TBS for 10 minutes at room temperature and blocked with 10% negative goat serum for 15 minutes at room temperature. Cells were probed with DRD1 Recombinant Monoclonal Antibody (Product # MA5-42467) at a dilution of 1:50 for 1 hour at room temperature, washed with PBS. Alexa Fluor 594 conjugate-Goat anti-Rabbit IgG was used as the secondary antibody at 1:1,000 dilution. The nuclear counter stai... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DRD1 Antibody in Immunocytochemistry (ICC/IF)
          DRD1 Antibody in Immunocytochemistry (ICC/IF)
          DRD1 Antibody in Immunocytochemistry (ICC/IF)
          DRD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DRD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DRD1 Antibody in Western Blot (WB)
          DRD1 Recombinant Rabbit Monoclonal Antibody (JM10-93)

          Product Details

          MA5-42467

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          JM10-93

          Immunogen

          Synthetic peptide within C-terminal human DRD1.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4, with 40% glycerol, 0.05% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2911608

          Product Specific Information

          Positive Control: Mouse kidney tissue lysates, 293T, N2A, SH-SY5Y, mouse brain tissue, rat brain tissue.

          Subcellular Location: Cell membrane. Endoplasmic reticulum membrane.

          Target Information

          The D1 subtype is the most abundant dopamine receptor in the central nervous system. This G-protein coupled receptor stimulates adenylyl cyclase and activates cyclic AMP-dependent protein kinases. D1 receptors regulate neuronal growth and development, mediate some behavioral responses, and modulate dopamine receptor D2-mediated events. Alternate transcription initiation sites result in two transcript variants of this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: D(1A) dopamine receptor; D1 receptor; D1A dopamine receptor; D1a receptor; Dopamine 1 receptor; Dopamine D1 receptor; dopamine receptor D1A; Dopamine-1A receptor; DRD1; G protein-coupled receptor DRD1; G-protein coupled receptor; unnamed protein product

          View more View less

          Gene Aliases: C030036C15Rik; D1a; D1R; DADR; Drd-1; DRD1; DRD1A; Gpcr15

          View more View less

          UniProt ID: (Human) P21728, (Mouse) Q61616

          View more View less

          Entrez Gene ID: (Human) 1812, (Mouse) 13488, (Rat) 24316

          View more View less

          Function(s)
          dopamine neurotransmitter receptor activity, coupled via Gs G-protein alpha-subunit binding G-protein coupled receptor activity dopamine neurotransmitter receptor activity protein binding heterotrimeric G-protein binding dopamine binding arrestin family protein binding signal transducer activity receptor binding drug binding G-protein coupled amine receptor activity protein phosphatase binding angiotensin receptor binding D3 dopamine receptor binding protein complex binding protein heterodimerization activity ATPase binding G-protein coupled receptor
          Process(es)
          temperature homeostasis conditioned taste aversion behavioral fear response neuron migration synaptic transmission, dopaminergic response to amphetamine protein import into nucleus muscle contraction signal transduction G-protein coupled receptor signaling pathway G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger adenylate cyclase-activating G-protein coupled receptor signaling pathway adenylate cyclase-activating dopamine receptor signaling pathway dopamine receptor signaling pathway synapse assembly learning memory mating behavior grooming behavior locomotory behavior adult walking behavior feeding behavior associative learning visual learning response to xenobiotic stimulus astrocyte development dopamine transport cell migration transmission of nerve impulse neuronal action potential dentate gyrus development striatum development hippocampus development cerebral cortex GABAergic interneuron migration positive regulation of cell migration peristalsis operant conditioning synaptic transmission, glutamatergic regulation of dopamine metabolic process response to cocaine vasodilation dopamine metabolic process maternal behavior positive regulation of potassium ion transport positive regulation of MAPK cascade glucose import habituation sensitization behavioral response to cocaine positive regulation of release of sequestered calcium ion into cytosol regulation of dopamine uptake involved in synaptic transmission positive regulation of synaptic transmission, glutamatergic prepulse inhibition phospholipase C-activating dopamine receptor signaling pathway long-term synaptic potentiation long term synaptic depression cellular response to catecholamine stimulus adenylate cyclase-activating adrenergic receptor signaling pathway modification of postsynaptic structure presynaptic modulation of chemical synaptic transmission positive regulation of neuron migration startle response negative regulation of protein kinase activity intracellular protein transport activation of adenylate cyclase activity chemical synaptic transmission sensory perception of chemical stimulus positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway regulation of vasoconstriction calcium-mediated signaling negative regulation of cell migration positive regulation of cAMP biosynthetic process phosphatidylinositol catabolic process social behavior negative regulation of circadian sleep/wake cycle, sleep response to drug regulation of ion transport positive regulation of adenylate cyclase activity positive regulation of membrane potential phosphatidylinositol metabolic process regulation of long-term neuronal synaptic plasticity positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway positive regulation of long-term synaptic potentiation positive regulation of feeding behavior response to organonitrogen compound response to organic cyclic compound response to activity orbitofrontal cortex development response to food response to estradiol response to retinoic acid cellular response to insulin stimulus response to nicotine response to antidepressant response to amino acid response to morphine response to ethanol response to steroid hormone cellular response to hypoxia
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline