Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • SR Antibodies

          Invitrogen

          SR Recombinant Rabbit Monoclonal Antibody (JE61-82)

          View all (3) SR antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SR Recombinant Rabbit Monoclonal Antibody (JE61-82)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (11)
          SR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          SR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SR Antibody (MA5-44854) in IHC (P)

          Immunohistochemistry analysis of SR protein repeat in paraffin-embedded human breast carcinoma tissue. The section was pre-treated using heat mediated antigen retrieval with sodium citrate buffer (pH 6.0) for 20 minutes. The tissues were blocked in 5% BSA for 30 minutes at room temperature, washed with ddH2O and PBS, and then probed with SR protein repeat Monoclonal antibody (Product # MA5-44854) using a di... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SR Antibody in Western Blot (WB)
          SR Recombinant Rabbit Monoclonal Antibody (JE61-82)

          Product Details

          MA5-44854

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          JE61-82

          Immunogen

          Synthetic peptide within human SR protein repeat aa 150-199/344.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4, with 0.05% BSA, 40% glycerol

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2931310

          Target Information

          SR proteins are a highly conserved family of arginine/serine-rich, spliceosome-associated phosphoproteins essential for metazoan pre-mRNA splicing. SR proteins act early in splicing by promoting splice site recognition and spliceosome assembly. SR proteins also play a regulatory role because they can determine alternative splice site usage in vivo and in vitro. SR proteins appear to be recruited from nuclear 'speckles', in which they are concentrated to sites of transcription in order to spatially coordinate transcription and pre-mRNA splicing within the cell nucleus.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Delayed-early protein HRS; epididymis secretory protein Li 91; epididymis secretory sperm binding protein; Insulin-induced growth response protein CL-4; pre-mRNA splicing factor SRp20; Pre-mRNA-splicing factor SRP20; Pre-mRNA-splicing factor SRP40; Pre-mRNA-splicing factor SRP55; Pre-mRNA-splicing factor SRP75; Protein X16; RNA binding protein; Serine/arginine-rich splicing factor 3; Serine/arginine-rich splicing factor 4; Serine/arginine-rich splicing factor 5; Serine/arginine-rich splicing factor 6; SFRS5; splicing factor arginine/serine-rich 5 (SRp40 HRS); Splicing factor, arginine/serine-rich 3; splicing factor, arginine/serine-rich 3 (SRp20); Splicing factor, arginine/serine-rich 4; splicing factor, arginine/serine-rich 4 (SRp75); Splicing factor, arginine/serine-rich 5; splicing factor, arginine/serine-rich 5 (SRp40, HRS); Splicing factor, arginine/serine-rich 6; splicing factor, arginine/serine-rich, 20-kD; splicing factor, arginine/serine-rich, 55 kDa; SR splicing factor 4; SR splicing factor 5; SR splicing factor 6; SRP001LB; unnamed protein product

          View more View less

          Gene Aliases: 1210001E11Rik; 5730499P16Rik; AI314910; AL024116; AW146126; AW550192; B52; Cl-4; HEL-S-91; HRS; MNCb-2616; RGD1561347; SFRS3; SFRS4; SFRS5; SFRS6; SRP20; SRP40; SRP55; SRP75; SRSF3; SRSF4; SRSF5; SRSF6; X16

          View more View less

          UniProt ID: (Human) P84103, (Human) Q08170, (Human) Q13243, (Human) Q13247, (Mouse) P84104, (Rat) Q09167, (Mouse) O35326, (Rat) G3V6S8, (Mouse) Q3TWW8

          View more View less

          Entrez Gene ID: (Human) 6428, (Human) 6429, (Human) 6430, (Human) 6431, (Rat) 361814, (Mouse) 20383, (Rat) 362612, (Mouse) 57317, (Rat) 29667, (Mouse) 20384, (Rat) 362264, (Mouse) 67996

          View more View less

          Function(s)
          nucleic acid binding RNA binding mRNA binding protein binding pre-mRNA binding phospholipase binding primary miRNA binding protein-RNA sequence-specific adaptor activity sequence-specific mRNA binding protein kinase B binding nucleotide binding poly(A) RNA binding RS domain binding RNA splicing factor
          Process(es)
          regulation of alternative mRNA splicing, via spliceosome RNA processing mRNA processing mRNA export from nucleus RNA splicing primary miRNA processing regulation of mRNA splicing, via spliceosome mRNA transport cellular response to leukemia inhibitory factor RNA splicing, via transesterification reactions response to insulin negative regulation of mRNA splicing, via spliceosome liver development mRNA splice site selection cellular response to insulin stimulus positive regulation of RNA splicing liver regeneration alternative mRNA splicing, via spliceosome negative regulation of gene expression regulation of keratinocyte proliferation negative regulation of keratinocyte differentiation positive regulation of epithelial cell proliferation involved in lung morphogenesis regulation of wound healing negative regulation of type B pancreatic cell apoptotic process transport insulin receptor signaling pathway hematopoietic progenitor cell differentiation transcription, DNA-templated regulation of transcription, DNA-templated response to wounding regulation of cell cycle negative regulation of cell death
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline