Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • VPS35 Antibodies

          Invitrogen

          VPS35 Monoclonal Antibody (11H10), APC

          View all (31) VPS35 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite VPS35 Monoclonal Antibody (11H10), APC

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (7)
          VPS35 Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          VPS35 Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          VPS35 Antibody (MA5-45948) in IHC

          Immunohistochemistry analysis using Intestinal Cancer. Samples were incubated with VPS35 monoclonal antibody (Product # MA5-45948) at 1:100 for Overnight at 4C, then 30 min at 37C, followed by Goat Anti-Mouse IgG (H+L): FITC for 45 min at 37C. Counterstain used was DAPI for 3 min at RT. Magnification: 20X. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          VPS35 Antibody in Immunohistochemistry (IHC)
          VPS35 Antibody in Western Blot (WB)
          VPS35 Antibody in Western Blot (WB)
          VPS35 Antibody in Immunoprecipitation (IP)
          VPS35 Antibody in Immunoprecipitation (IP)
          VPS35 Antibody in Immunoprecipitation (IP)
          VPS35 Antibody in Immunoprecipitation (IP)
          VPS35 Monoclonal Antibody (11H10), APC

          Product Details

          MA5-45948

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (IHC)

          1:100
          -

          Immunoprecipitation (IP)

          1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG2b

          Class

          Monoclonal

          Type

          Antibody

          Clone

          11H10

          Immunogen

          Full length recombinant human VSP35
          3D Epitope / Immunogen

          Conjugate

          APC APC APC
          • FITC
          • PerCP 
          • Request custom conjugation

          Excitation/Emission Max

          651/660 nm View spectra spectra

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          95.64mM phosphate/2.48mM MES, pH 7.4, with 0.5M EDTA

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2932402

          Product Specific Information

          A 1:1,000 dilution of MA5-45948 was sufficient for detection of VPS35 in 10 µg of SH-SY5Y by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary antibody.|Detects approximately 92 kDa.

          Target Information

          This gene belongs to a group of vacuolar protein sorting (VPS) genes. The encoded protein is a component of a large multimeric complex, termed the retromer complex, involved in retrograde transport of proteins from endosomes to the trans-Golgi network. The close structural similarity between the yeast and human proteins that make up this complex suggests a similarity in function. Expression studies in yeast and mammalian cells indicate that this protein interacts directly with VPS35, which serves as the core of the retromer complex.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: DKFZp434E1211; DKFZp434P1672; FLJ10752; FLJ13588; FLJ20388; hVPS35; Maternal embryonic message 3; Maternal-embryonic 3; MEM3; similar to Homo sapiens vacuolar sorting protein 35 (VPS35) mRNA with GenBank Accession Number AF191298; unnamed protein product; vacuolar protein sorting 35 homolog; Vacuolar protein sorting-associated protein 35; Vesicle protein sorting 35; VPS35

          View more View less

          Gene Aliases: AI647796; MEM3; PARK17; TCCCTA00141; VPS35

          View more View less

          UniProt ID: (Human) Q96QK1, (Mouse) Q9EQH3

          View more View less

          Entrez Gene ID: (Human) 55737, (Rat) 25479, (Mouse) 65114

          View more View less

          Function(s)
          protein binding D1 dopamine receptor binding protein transporter activity membrane traffic protein
          Process(es)
          intracellular protein transport lysosome organization positive regulation of signal transduction positive regulation of gene expression negative regulation of gene expression regulation of mitochondrion organization protein transport Wnt signaling pathway regulation of macroautophagy regulation of protein stability protein destabilization endocytic recycling negative regulation of protein homooligomerization protein localization to organelle protein localization to endosome retrograde transport, endosome to Golgi regulation of protein catabolic process mitochondrial fragmentation involved in apoptotic process transcytosis positive regulation of protein catabolic process negative regulation of inflammatory response regulation of developmental process modulation of synaptic transmission voluntary musculoskeletal movement regulation of transport regulation of protein metabolic process positive regulation of dopamine receptor signaling pathway positive regulation of Wnt protein secretion regulation of synapse maturation positive regulation of mitochondrial fission positive regulation of canonical Wnt signaling pathway positive regulation of locomotion involved in locomotory behavior neurotransmitter receptor transport, endosome to postsynaptic membrane vesicle-mediated transport in synapse mitochondrion to lysosome transport regulation of postsynapse organization neurotransmitter receptor transport, endosome to plasma membrane regulation of postsynapse assembly negative regulation of late endosome to lysosome transport regulation of dendritic spine maintenance positive regulation of dopamine biosynthetic process negative regulation of cellular protein localization positive regulation of cellular protein localization positive regulation of protein localization to cell periphery negative regulation of lysosomal protein catabolic process regulation of presynapse assembly regulation of terminal button organization retrograde transport, endosome to plasma membrane vacuolar protein processing transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline