Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • ZC3H12A Antibodies

          Invitrogen

          ZC3H12A Recombinant Rabbit Monoclonal Antibody (HL1442)

          View all (8) ZC3H12A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ZC3H12A Recombinant Rabbit Monoclonal Antibody (HL1442)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          ZC3H12A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ZC3H12A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ZC3H12A Antibody (MA5-47045) in WB

          Western blot analysis of ZC3H12A in 293T whole cell extracts (30 µg) separated by 7.5% SDS-PAGE. Membrane was blotted with ZC3H12A Monoclonal antibody (Product # MA5-47045) using a dilution of 1:1,000 followed by HRP-conjugated anti-rabbit IgG secondary antibody. The signal was developed with Trident ECL plus-Enhanced. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ZC3H12A Antibody in Western Blot (WB)
          ZC3H12A Antibody in Western Blot (WB)
          ZC3H12A Antibody in Western Blot (WB)

          Product Details

          MA5-47045

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          -
          Product Specifications

          Species Reactivity

          Cat, Human

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          HL1442

          Immunogen

          Recombinant protein encompassing a sequence within the center region of human ZC3H12A. The exact sequence is proprietary.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2938117

          Product Specific Information

          Store as concentrated solution.

          Centrifuge briefly prior to opening vial.

          Target Information

          ZC3H12A is an MCP1 -induced protein that acts as a transcriptional activator and causes cell death of cardiomyocytes, possibly via induction of genes associated with apoptosis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: bifunctional endoribonuclease and deubiquitinase ZC3H12A; Endoribonuclease ZC3H12A; FLJ23231; MCP-1 induced protein 1; MCPIP1; MCP-1 treatment-induced protein; MCP-induced protein 1; MCPIP; transcription factor; Monocyte chemotactic protein-induced protein 1; Reg1; Regnase-1; ribonuclease ZC3H12A; RP3-423B22.1; unnamed protein product; Zinc finger CCCH domain-containing protein 12A

          View more View less

          Gene Aliases: dJ423B22.1; MCPIP; MCPIP-1; MCPIP1; Reg1; ZC3H12A

          View more View less

          UniProt ID: (Human) Q5D1E8

          View more View less

          Entrez Gene ID: (Human) 80149

          View more View less

          Function(s)
          DNA binding chromatin binding RNA binding mRNA binding mRNA 3'-UTR binding nuclease activity endonuclease activity endoribonuclease activity exoribonuclease activity ribonuclease activity thiol-dependent ubiquitin-specific protease activity protein binding zinc ion binding hydrolase activity miRNA binding RNA stem-loop binding mRNA 3'-UTR AU-rich region binding ribosome binding metal ion binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter nuclear-transcribed mRNA catabolic process, nonsense-mediated decay angiogenesis negative regulation of protein phosphorylation positive regulation of defense response to virus by host immune system process immune response-activating signal transduction mRNA catabolic process apoptotic process inflammatory response cellular response to DNA damage stimulus nervous system development regulation of gene expression positive regulation of autophagy miRNA catabolic process positive regulation of endothelial cell migration positive regulation of gene expression negative regulation of gene expression negative regulation of muscle cell apoptotic process positive regulation of lipid storage protein deubiquitination cell differentiation negative regulation of interferon-gamma production negative regulation of interleukin-1 beta production negative regulation of interleukin-6 production negative regulation of tumor necrosis factor production cellular response to oxidative stress cellular response to glucose starvation positive regulation of protein import into nucleus negative regulation of macrophage activation negative regulation of I-kappaB kinase/NF-kappaB signaling negative regulation by host of viral genome replication negative regulation of nitric oxide biosynthetic process positive regulation of fat cell differentiation positive regulation of angiogenesis positive regulation of transcription from RNA polymerase II promoter T cell receptor signaling pathway protein oligomerization defense response to virus negative regulation of cardiac muscle contraction positive regulation of mRNA catabolic process 3'-UTR-mediated mRNA destabilization cellular response to lipopolysaccharide cellular response to interleukin-1 cellular response to tumor necrosis factor cellular response to virus negative regulation of cytokine production involved in inflammatory response positive regulation of execution phase of apoptosis positive regulation of p38MAPK cascade negative regulation of NIK/NF-kappaB signaling positive regulation of protein deubiquitination cellular response to sodium arsenite cellular response to ionomycin cellular response to chemokine negative regulation of T-helper 17 cell differentiation positive regulation of reactive oxygen species metabolic process positive regulation of miRNA catabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline