Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • PIP5K1A Antibodies

          Invitrogen

          PIP5K1A Recombinant Rabbit Monoclonal Antibody (HL2011)

          View all (12) PIP5K1A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PIP5K1A Recombinant Rabbit Monoclonal Antibody (HL2011)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          PIP5K1A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PIP5K1A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PIP5K1A Antibody (MA5-47257) in ICC/IF

          Immunofluorescent analysis of PIP5K1A in HeLa cells. Cells were fixed in 4% paraformaldehyde at RT for 15 min. Green: PIP5K1A stained by PIP5K1A monoclonal antibody (Product # MA5-47257) diluted at 1:500. Blue: Fluoroshield with DAPI. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PIP5K1A Antibody in Immunocytochemistry (ICC/IF)
          PIP5K1A Antibody in Western Blot (WB)
          PIP5K1A Antibody in Western Blot (WB)
          PIP5K1A Antibody in Western Blot (WB)
          PIP5K1A Recombinant Rabbit Monoclonal Antibody (HL2011)

          Product Details

          MA5-47257

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Dog, Cat, Human, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          HL2011

          Immunogen

          Recombinant protein encompassing a sequence within the N-terminus region of human PIP5K1A. The exact sequence is proprietary.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2938329

          Product Specific Information

          Store as concentrated solution.

          Centrifuge briefly prior to opening vial.

          Target Information

          PIP5K1A lipid kinase or phosphatidylinositol-4-phosphate 5-kinase 1 alpha phosphorylates phosphatidylinositol-4-phosphate to form phosphatidylinositol 4,5-bisphosphate. PIP5K1 is required for Wnt3a-induced LRP6 phosphorylation at ser1490 in mammalian cells. Wnt3a stimulates the formation of phosphatidylinositol 4,5-bisphosphates through 'frizzled' and 'disheveled', the latter of which directly interacts with and activated PIP5KI. PIP5K1A-mediated PIP(2) production is crucial for HIV-1 entry and the early steps of infection in permissive lymphocytes. Overexpression of wild-type PI4P5K1A increases HIV-1 Env-mediated PIP(2) production and enhances viral replication in primary lymphocytes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 68 kDa type I phosphatidylinositol 4-phosphate 5-kinase; 68 kDa type I phosphatidylinositol 4-phosphate 5-kinase alpha; Phosphatidylinositol 4-phosphate 5-kinase type I alpha; Phosphatidylinositol 4-phosphate 5-kinase type-1 alpha; phosphatidylinositol-4-phosphate 5-kinase; phosphatidylinositol-4-phosphate 5-kinase type-1 alpha; phosphatidylinositol-4-phosphate 5-kinase, type 1 beta; phosphatidylinositol-4-phosphate 5-kinase, type I, alpha; PI51A; PIP5K1-alpha; PIP5KIalpha; ptdIns(4)P-5-kinase 1 alpha; unnamed protein product

          View more View less

          Gene Aliases: PIP5K1A; Pip5k1b

          View more View less

          UniProt ID: (Human) Q99755, (Rat) D3ZSI8

          View more View less

          Entrez Gene ID: (Human) 8394, (Dog) 475846, (Rat) 365865

          View more View less

          Function(s)
          nucleotide binding 1-phosphatidylinositol-3-phosphate 5-kinase activity protein binding ATP binding kinase activity 1-phosphatidylinositol-4-phosphate 5-kinase activity transferase activity kinase binding phosphatidylinositol kinase activity 1-phosphatidylinositol-5-kinase activity kinase
          Process(es)
          lipid metabolic process glycerophospholipid metabolic process phosphatidylinositol biosynthetic process phagocytosis signal transduction phospholipid biosynthetic process fibroblast migration cell migration actin cytoskeleton organization keratinocyte differentiation phosphatidylinositol metabolic process phosphatidylinositol phosphorylation focal adhesion assembly regulation of protein kinase B signaling cell chemotaxis protein localization to plasma membrane activation of GTPase activity ruffle assembly actin cytoskeleton reorganization protein targeting to plasma membrane
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline