Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Aurora A/B Antibodies

          Invitrogen

          Aurora A/B Monoclonal Antibody (3H1)

          View all (2) Aurora A/B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Aurora A/B Monoclonal Antibody (3H1)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          Aurora A/B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Aurora A/B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Aurora A/B Antibody (MA5-47418) in ICC/IF

          Immunofluorescent analysis of Aurora A in HeLa cell culture. Samples were stained with Aurora A monoclonal antibody (Product # MA5-47418) in green. Costained with chicken pAb to vimentin in red. The blue is DAPI staining of nuclear DNA. The aurora A antibody stains spindle poles and mitotic spindles at anaphase and concentrates on the midbody between the two daughter cells during telophase. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Aurora A/B Antibody in Immunocytochemistry (ICC/IF)
          Aurora A/B Antibody in Western Blot (WB)
          Aurora A/B Monoclonal Antibody (3H1)

          Product Details

          MA5-47418

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (IHC)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          3H1

          Immunogen

          Full length recombinant human Aurora A protein expressed in and purified from E. coli.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A/G

          Storage buffer

          PBS with 50% glycerol

          Contains

          5mM sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2942400

          Target Information

          The Aurora gene, a serine-threonine kinase, orginally discovered in Drosophila, has been found to be responsible for a chromosomal segregation defect. The mammalian homologues of Drosophila Aurora have been termed ARK1 (Aurora-related kinase 1) and ARK2.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: AIM-1; ARK-1; ARK-2; Aurora 1; Aurora 2; aurora A; aurora B; aurora family kinase 1; Aurora kinase A; Aurora kinase B; aurora kinase B-Sv1; aurora kinase B-Sv2; Aurora- and IPL1-like midbody-associated protein 1; aurora- and Ipl1-like midbody-associated protein 1 homolog; aurora-1; aurora-A; aurora-B; aurora-related kinase 1; aurora-related kinase 2; aurora/IPL1-like kinase; Aurora/IPL1-related kinase 1; Aurora/IPL1-related kinase 2; Breast tumor-amplified kinase; Ipl1- and aurora-related kinase 1; protein phosphatase 1, regulatory subunit 47; protein phosphatase 1, regulatory subunit 48; ratAurA; serine/threonine kinase 12; serine/threonine kinase 5; serine/threonine kinase 6; serine/threonine protein kinase 15; serine/threonine protein kinase 6; Serine/threonine-protein kinase 12; Serine/threonine-protein kinase 15; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase 6; Serine/threonine-protein kinase aurora-A; Serine/threonine-protein kinase aurora-B; Serine/threonine-protein kinase Ayk1; STK-1; unnamed protein product

          View more View less

          Gene Aliases: AIK; AIK2; AIM-1; AIM1; AIRK1; AIRK2; AL022959; ARK-1; ARK-2; ARK1; ARK2; AU019385; AURA; AurB; AURKA; AURKB; aurkb-sv1; aurkb-sv2; Aurora-A; AW539821; AYK1; BTAK; IAK; IAK1; IPL1; PPP1R47; PPP1R48; STK-1; STK1; STK12; STK15; STK5; STK6; STK7

          View more View less

          UniProt ID: (Human) O14965, (Human) Q96GD4, (Mouse) P97477, (Mouse) O70126, (Rat) O55099

          View more View less

          Entrez Gene ID: (Human) 6790, (Human) 9212, (Mouse) 20878, (Rat) 261730, (Mouse) 20877, (Rat) 114592

          View more View less

          Function(s)
          nucleotide binding protein kinase activity protein serine/threonine kinase activity protein serine/threonine/tyrosine kinase activity protein binding ATP binding kinase activity transferase activity protein kinase binding ubiquitin protein ligase binding histone kinase activity (H3-S10 specific) protein heterodimerization activity protein serine kinase activity molecular function activator activity kinase binding transferase activity, transferring phosphorus-containing groups histone serine kinase activity metal ion binding non-receptor serine/threonine protein kinase
          Process(es)
          G2/M transition of mitotic cell cycle meiotic spindle organization microtubule cytoskeleton organization mitotic cell cycle chromatin remodeling protein phosphorylation apoptotic process spindle organization mitotic spindle organization spindle assembly involved in female meiosis I centrosome cycle mitotic centrosome separation response to wounding anterior/posterior axis specification regulation of G2/M transition of mitotic cell cycle negative regulation of gene expression peptidyl-serine phosphorylation cell projection organization regulation of protein stability negative regulation of protein binding positive regulation of proteasomal ubiquitin-dependent protein catabolic process regulation of cytokinesis regulation of microtubule-based process negative regulation of apoptotic process proteasome-mediated ubiquitin-dependent protein catabolic process cell cycle G2/M phase transition positive regulation of mitotic nuclear division positive regulation of mitotic cell cycle regulation of centrosome cycle protein autophosphorylation cell division meiotic cell cycle centrosome localization cilium disassembly protein localization to centrosome positive regulation of cell cycle process positive regulation of mitochondrial fission liver regeneration positive regulation of oocyte maturation regulation of signal transduction by p53 class mediator neuron projection extension negative regulation of transcription from RNA polymerase II promoter mitotic cytokinesis negative regulation of B cell apoptotic process positive regulation of microtubule depolymerization positive regulation of telomere maintenance negative regulation of cytokinesis positive regulation of cytokinesis protein localization to kinetochore cellular response to UV cleavage furrow formation post-translational protein modification mitotic cytokinesis checkpoint negative regulation of innate immune response mitotic spindle midzone assembly attachment of mitotic spindle microtubules to kinetochore regulation of chromosome segregation midbody abscission positive regulation of mitotic sister chromatid segregation positive regulation of mitotic cell cycle spindle assembly checkpoint mitotic spindle assembly repair of mitotic kinetochore microtubule attachment defect cGAS/STING signaling pathway negative regulation of cGAS/STING signaling pathway positive regulation of mitotic sister chromatid separation positive regulation of attachment of mitotic spindle microtubules to kinetochore positive regulation of mitotic cytokinesis positive regulation of lateral attachment of mitotic spindle microtubules to kinetochore mitotic sister chromatid biorientation cell cycle mitotic nuclear division meiotic nuclear division phosphorylation histone-serine phosphorylation centrosome organization negative regulation of spindle checkpoint response to estradiol spindle checkpoint histone H3-S28 phosphorylation aging cell proliferation abscission
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline