Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Rhodopsin Antibodies

          Invitrogen

          Rhodopsin Monoclonal Antibody (B630)

          View all (30) Rhodopsin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Rhodopsin Monoclonal Antibody (B630)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          Rhodopsin Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          Rhodopsin Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Rhodopsin Antibody (MA5-47474) in IHC

          Immunohistochemistry analysis of Rhodopsin in mouse retina section. Samples were stained with Rhodopsin monoclonal antibody (Product # MA5-47474) using a dilution of 1:2,000 (green). Costained with rabbit pAb to Fox3/NeuN (Product # PA5-143605) dilution 1:5,000 in red. The blue is Hoechst staining of nuclear DNA. Rhodopsin antibody reveals the rod cell membranes located in photoreceptor outer segments (OS) layer of the retina. The Fox3/NeuN antibody selectively stains the nuclei and cytoplasm of neuronal cells in the ganglion cell layer (G... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Rhodopsin Antibody in Immunohistochemistry (IHC)
          Rhodopsin Antibody in Immunohistochemistry (IHC)
          Rhodopsin Antibody in Western Blot (WB)
          Rhodopsin Monoclonal Antibody (B630)

          Product Details

          MA5-47474

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000
          -

          Immunohistochemistry (IHC)

          1:2,000
          -

          Immunocytochemistry (ICC/IF)

          1:2,000
          -
          Product Specifications

          Species Reactivity

          Bovine, Horse, Human, Mouse, Pig, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          B630

          Immunogen

          Purified bovine rhodopsin
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          PBS with 50% glycerol

          Contains

          5mM sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2942456

          Target Information

          Visual pigments such as rhodopsin and porphyropsin are light-absorbing molecules that mediate vision. Rhodopsin consists of an apoprotein, opsin, covalently linked to 11-cis-retinal. This receptor is coupled to the activation of phospholipase C. Porphyropsin consists of opsin covalently linked to 11-cis 3,4-didehydroretinal.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: gamma-aminobutyric acid receptor subunit rho1; L opsin; Long Wavelength Sensitive opsin; LWS opsin; MGC138309; MGC138311; OPN 2; opsin 2; opsin 2, rod pigment; Opsin-2; Red Opsin; Rhodopsin; rhodopsin (opsin 2, rod pigment) (retinitis pigmentosa 4, autosomal dominant); Rhodopsin (retinitis pigmentosa 4, autosomal dominant); Rod Opsin; rod visual pigment; RP 4

          View more View less

          Gene Aliases: CSNBAD1; Noerg1; OPN2; Ops; PIGRHO1; RHO; RHO1; RP4

          View more View less

          UniProt ID: (Bovine) P02699, (Human) P08100, (Pig) O18766, (Rat) P51489, (Mouse) P15409

          View more View less

          Entrez Gene ID: (Bovine) 509933, (Human) 6010, (Pig) 397437, (Horse) 100056641, (Rat) 24717, (Mouse) 212541

          View more View less

          Function(s)
          G-protein alpha-subunit binding opsin binding guanyl-nucleotide exchange factor activity 11-cis retinal binding protein binding G-protein coupled photoreceptor activity zinc ion binding identical protein binding arrestin family protein binding G-protein coupled receptor activity photoreceptor activity metal ion binding retinal binding spectrin binding signal transducer activity G-protein coupled receptor
          Process(es)
          protein phosphorylation visual perception phototransduction, visible light response to light stimulus absorption of visible light rhodopsin mediated signaling pathway protein-chromophore linkage photoreceptor cell maintenance retina development in camera-type eye microtubule cytoskeleton organization signal transduction G-protein coupled receptor signaling pathway phototransduction detection of light stimulus response to light intensity gene expression thermotaxis sensory perception of light stimulus detection of temperature stimulus involved in thermoception cellular response to light stimulus podosome assembly rod bipolar cell differentiation red, far-red light phototransduction response to stimulus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline