Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
G6D, also known as Lymphocyte antigen 6 complex locus G6D (LY6G6D), is a type I transmembrane protein belonging to the immunoglobin (Ig) superfamily, that is comprised of cell-surface proteins involved in the immune system and cellular recognition. It is a disulfide linked homodimer with an Ig-like V-type domain and is known to interact with GRB2 and GRB7 in a phosphorylation-dependent manner thus playing an important role in their downstream signal transduction pathways.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DADB-110M10.6; lymphocyte antigen 6 complex G6D; Lymphocyte antigen 6 complex locus protein G6d; lymphocyte antigen 6 complex, locus G6D; lymphocyte antigen 6 complex, locus G6F; lymphocyte antigen-6 G6D; megakaryocyte-enhanced gene transcript 1; Megakaryocyte-enhanced gene transcript 1 protein; Protein Ly6-D
Gene Aliases: A930024F17Rik; C6orf23; G6D; G6f; LY6-D; LY6G6D; LY6G6F; MEGT1; NG25; NG32
UniProt ID: (Human) O95868, (Rat) Q6MG58, (Mouse) Q9Z1Q3
Entrez Gene ID: (Cynomolgus monkey) 102119090, (Human) 58530, (Rat) 415062, (Mouse) 114654
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support