Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GNAI1/GNAI2/GNAI3 Antibodies

          Invitrogen

          GNAI1/GNAI2/GNAI3 Recombinant Rabbit Monoclonal Antibody (HL2092)

          View all (2) GNAI1/GNAI2/GNAI3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite GNAI1/GNAI2/GNAI3 Recombinant Rabbit Monoclonal Antibody (HL2092)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          GNAI1/GNAI2/GNAI3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          GNAI1/GNAI2/GNAI3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GNAI1/GNAI2/GNAI3 Antibody (MA5-50378) in IHC (P)

          Immunohistochemical analysis of GNAI1/GNAI2/GNAI3 using GNAI1/GNAI2/GNAI3 Recombinant Monoclonal Antibody (Product # MA5-50378) in Paraffin embedded rat duodenum and kidney tissue at a dilution of 1:100. For Antigen retrieval citrate buffer, pH 6.0, as used for 15 min. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GNAI1/GNAI2/GNAI3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GNAI1/GNAI2/GNAI3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GNAI1/GNAI2/GNAI3 Antibody in Western Blot (WB)
          GNAI1/GNAI2/GNAI3 Antibody in Western Blot (WB)
          GNAI1/GNAI2/GNAI3 Antibody in Western Blot (WB)
          GNAI1/GNAI2/GNAI3 Antibody in Western Blot (WB)

          Product Details

          MA5-50378

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat, Zebrafish

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          HL2092

          Immunogen

          Recombinant fragment of human GNAI3.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3094420

          Target Information

          GNAI1/GNAI2/GNAI3 are genes that encode for the G protein alpha inhibiting activity subunit. These proteins play a crucial role in signal transduction pathways, particularly in the inhibition of adenylyl cyclase. They are involved in various physiological processes, including neurotransmitter release, hormone secretion, and regulation of smooth muscle contraction. Mutations in these genes have been associated with various diseases, including hypoparathyroidism, pituitary adenoma, and cardiovascular disorders.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Adenylate cyclase-inhibiting G alpha protein; G alpha inhibitory type 2 (a); G alpha inhibitory type 2 (b); G protein; G protein alpha subunit; G(i) alpha-3; Galpha i1a; Galpha i3; Gi1 protein alpha subunit; GNAI2; GTP-binding protein (G-alpha-i2); GTP-binding protein alpha subunit; GTP-binding regulatory protein Gi alpha-2 chain; guanine nucleotide binding protein (G protein), alpha inhibiting 1; guanine nucleotide binding protein (G protein), alpha inhibiting 2; guanine nucleotide binding protein (G protein), alpha inhibiting 3; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 1; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 2; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 3; guanine nucleotide binding protein, alpha inhibiting 1; guanine nucleotide binding protein, alpha inhibiting 2; guanine nucleotide binding protein, alpha inhibiting 3; Guanine nucleotide binding, protein, alpha inhibiting polypeptide 3; guanine nucleotide-binding protein (G) alpha subunit; guanine nucleotide-binding protein alpha-i subunit; guanine nucleotide-binding protein G(i) subunit alpha; Guanine nucleotide-binding protein G(i) subunit alpha-1; Guanine nucleotide-binding protein G(i) subunit alpha-2; Guanine nucleotide-binding protein G(i) subunit alpha-3; guanine nucleotide-binding protein G(i), alpha-1 subunit; guanine nucleotide-binding protein G(i), alpha-2 subunit; guanine nucleotide-binding protein G(k) subunit alpha; guanine nucleotide-binding regulatory protein alpha-inhibitory subunit; heterotrimeric guanine nucleotide-binding protein 1A; heterotrimeric guanine nucleotide-binding protein 1B; heterotrimeric guanine nucleotide-binding protein 1C; HG1B/HG1C/HG1A; sGi2; unnamed protein product

          View more View less

          Gene Aliases: 87U6; AI158965; ARCND1; ARCODS; AU046200; AW537698; BPGTPB; C76432; Galphai2; Galphai3; Gi; Gia; Gialpha1; GIP; GIP3A; Gnai-1; Gnai-2; Gnai-3; GNAI1; GNAI2; GNAI2B; GNAI3; H_LUCA15.1; H_LUCA16.1; HG1A; HG1B; HG1C; NEDHISB; zgc:63957

          View more View less

          UniProt ID: (Human) P63096, (Human) P04899, (Human) P08754, (Rat) P10824, (Mouse) B2RSH2, (Rat) P04897, (Mouse) P08752, (Rat) P08753, (Mouse) Q9DC51

          View more View less

          Entrez Gene ID: (Human) 2770, (Human) 2771, (Human) 2773, (Rat) 25686, (Mouse) 14677, (Rat) 81664, (Mouse) 14678, (Rat) 25643, (Mouse) 14679, (Zebrafish) 393946, (Zebrafish) 100006888

          View more View less

          Function(s)
          nucleotide binding magnesium ion binding G-protein coupled receptor binding GTPase activity obsolete small monomeric GTPase activity protein binding GTP binding adenylate cyclase regulator activity adenylate cyclase inhibitor activity hydrolase activity guanyl nucleotide binding GDP binding G-protein beta/gamma-subunit complex binding D2 dopamine receptor binding G-protein coupled serotonin receptor binding metal ion binding signal transducer activity GTPase activating protein binding protein domain specific binding heterotrimeric G-protein
          Process(es)
          signal transduction G-protein coupled receptor signaling pathway adenylate cyclase-modulating G-protein coupled receptor signaling pathway adenylate cyclase-activating G-protein coupled receptor signaling pathway adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway adenylate cyclase-inhibiting serotonin receptor signaling pathway neuropeptide signaling pathway response to prostaglandin E response to peptide hormone positive regulation of cholesterol biosynthetic process negative regulation of insulin secretion cell division regulation of mitotic spindle organization chemokine-mediated signaling pathway T cell migration positive regulation of relaxation of smooth muscle cellular response to forskolin positive regulation of protein localization to cell cortex adenosine receptor signaling pathway negative regulation of adenylate cyclase activity G-protein coupled acetylcholine receptor signaling pathway gamma-aminobutyric acid signaling pathway response to nutrient cell proliferation positive regulation of cell proliferation positive regulation of cell migration positive regulation of superoxide anion generation positive regulation of urine volume negative regulation of calcium ion-dependent exocytosis modulation of synaptic transmission negative regulation of synaptic transmission regulation of calcium ion transport positive regulation of ERK1 and ERK2 cascade negative regulation of adrenergic receptor signaling pathway positive regulation of vascular smooth muscle cell proliferation positive regulation of neural precursor cell proliferation negative regulation of apoptotic signaling pathway positive regulation of macroautophagy GTP metabolic process metabolic process cell cycle activation of MAPKK activity G-protein coupled glutamate receptor signaling pathway regulation of heart contraction intracellular signal transduction vesicle fusion negative regulation of secretion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline