Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Immunogen sequence: FENDSFLQRV IKTYKKIKKD SAMLLSGCSH LLHNKELMAS LAESSFDVML TDPFLPCSPI VAQYLSLPTV FFLHALPCSL EFEATQCPNP FSYVPRPLSS H
This gene encodes a UDP-glucuronosyltransferase, an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. This gene is part of a complex locus that encodes several UDP-glucuronosyltransferases. The locus includes thirteen unique alternate first exons followed by four common exons. Four of the alternate first exons are considered pseudogenes. Each of the remaining nine 5' exons may be spliced to the four common exons, resulting in nine proteins with different N-termini and identical C-termini. Each first exon encodes the substrate binding site, and is regulated by its own promoter. The preferred substrate of this enzyme is bilirubin, although it also has moderate activity with simple phenols, flavones, and C18 steroids. Mutations in this gene result in Crigler-Najjar syndromes types I and II and in Gilbert syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B1; EC 2.4.1.17; HUG BR1; HUGBR1; UDP glycosyltransferase 1 family, polypeptide A1; UDP-glucuronosyltransferase 1 family member 1; UDP-glucuronosyltransferase 1 family, member 1; UDP-glucuronosyltransferase 1 family, polypeptide A1; UDP-glucuronosyltransferase 1-1; UDP-glucuronosyltransferase 1A1; UDPGT; UDPGT 1-1; UGT 1A; UGT1 01; UGT1*1; UGT1-01; UGT1.1; UGT1A1; UGTBR1
Gene Aliases: Gnt1; Udpgt; UDPGT 1-1; Udpgt-1a; Ugt1; UGT1A01; Ugt1a1; UgtBr1
UniProt ID: (Mouse) Q63886, (Rat) Q64550
Entrez Gene ID: (Mouse) 394436, (Rat) 24861
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support