Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen Sequence: TTSTFC
PKC alpha/delta refers to the protein kinase C isoforms alpha and delta, which are gene targets involved in various cellular processes and signaling pathways. Protein kinase C (PKC) is a family of enzymes that play a crucial role in regulating cell growth, differentiation, and intracellular signaling. PKC alpha and delta isoforms are widely expressed in different tissues and have distinct functions. PKC alpha is involved in cell proliferation, apoptosis, and cell cycle regulation, while PKC delta is implicated in cellular responses to stress, DNA damage, and apoptosis. These isoforms are activated by various stimuli, including second messengers and receptor-mediated signaling, leading to the phosphorylation of downstream targets and modulation of cellular processes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AAG6; aging-associated gene 6; nPKC-delta; PKC-A; PKCA; PKCdeltaVIII; PRKACA; Protein kinase C; Protein kinase C alpha type; protein kinase C alpha-polypeptide; Protein kinase C delta type; protein kinase C delta VIII; Tyrosine-protein kinase PRKCD; unnamed protein product
Gene Aliases: AAG6; ALPS3; CVID9; MAY1; nPKC-delta; PKC-alpha; PKCA; PKCalpha; PKCD; PKCI+/-; PRKACA; PRKCA; PRKCD
UniProt ID: (Human) P17252, (Human) Q05655
Entrez Gene ID: (Human) 5578, (Human) 5580
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support