Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Immunogen Sequence: PLRAPGPLAA AVPADKARSP AVPLLLPPAL AETVGPAPPG VLRCALGDRG RVRGRAAPYC VAELATGPSA LSPLPPQADL DGPPGAGKQG IPQPLSGPCR R
The protein encoded by this gene belongs to the GSK-3-binding protein family. The protein inhibits GSK-3-mediated phosphorylation of beta-catenin and positively regulates the Wnt signaling pathway. It may function in tumor progression and in lymphomagenesis. [provided by RefSeq, Oct 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FRAT-1; FRAT1, WNT signaling pathway regulator; Frequently rearranged in advanced T-cell lymphomas 1; Proto-oncogene FRAT1
Gene Aliases: AW060382; FRAT1
UniProt ID: (Human) Q92837, (Mouse) P70339
Entrez Gene ID: (Human) 10023, (Rat) 103691373, (Mouse) 14296
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support