Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Immunogen Sequence: MEADGDGEEL ARLRSVFAAC DANRSGRLER EEFRALCTEL RVRPADAEAV FQRLDADRDG AITFQEFARG FLGSLRGGRR RDWGPLDPAP AVSEAGPETH DSEEDEGDED AAAALATSCG
Binds predominantly GDP, and also GTP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ras and EF-hand domain-containing protein; ras and EF-hand domain-containing protein homolog; Ras-related protein Rab-45; unnamed protein product
Gene Aliases: BC030882; RAB45; RASEF; TSG
UniProt ID: (Human) Q8IZ41, (Mouse) Q5RI75
Entrez Gene ID: (Human) 158158, (Mouse) 242505
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support