Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
This gene encodes the glycogen debrancher enzyme which is involved in glycogen degradation. This enzyme has two independent catalytic activities which occur at different sites on the protein: a 4-alpha-glucotransferase activity and a amylo-1,6-glucosidase activity. Mutations in this gene are associated with glycogen storage disease although a wide range of enzymatic and clinical variability occurs which may be due to tissue-specific alternative splicing. Alternatively spliced transcripts encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-Apr; 4-glucantransferase; amylo-1, 6-glucosidase, 4-alpha-glucanotransferase; amylo-1, 6-glucosidase, 4-alpha-glucanotransferase (glycogen debranching enzyme, glycogen storage disease type III); amylo-1,6-glucosidase, 4-alpha-glucanotransferase; amylo-alpha-1, 6-glucosidase, 4-alpha-glucanotransferase; amylo-alpha-1, 6-glucosidase, 4-alpha-glucanotransferaseprovided; Glycogen debrancher; Glycogen debranching enzyme; glycogen debranching protein; Oligo-1
Gene Aliases: 1110061O17Rik; 9430004C13Rik; 9630046L06Rik; AGL; AI850929; C77197; GDE
UniProt ID: (Human) P35573
Entrez Gene ID: (Human) 178, (Rat) 362029, (Mouse) 77559
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support