Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Enzyme that can either act as an ADP-sugar pyrophosphatase in absence of diphosphate or catalyze the synthesis of ATP in presence of diphosphate. In absence of diphosphate, hydrolyzes with similar activities various modified nucleoside diphosphates such as ADP-ribose, ADP-mannose, ADP-glucose, 8-oxo-GDP and 8-oxo-dGDP. Can also hydrolyze other nucleotide sugars with low activity. In presence of diphosphate, mediates the synthesis of ATP in the nucleus by catalyzing the conversion of ADP-ribose to ATP and ribose 5-phosphate. Nuclear ATP synthesis takes place when dephosphorylated at Thr-45. Nuclear ATP generation is required for extensive chromatin remodeling events that are energy-consuming. Does not play a role in U8 snoRNA decapping activity. Binds U8 snoRNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 8-oxo-dGDP phosphatase; ADP-sugar pyrophosphatase; Nuclear ATP-synthesis protein NUDIX5; Nucleoside diphosphate-linked moiety X motif 5; nudix (nucleoside diphosphate linked moiety X)-type motif 5; nudix hydrolase; Nudix motif 5; NUDT5; unnamed protein product; YSA1H
Gene Aliases: hNUDT5; HSPC115; NUDIX5; NUDT5; YSA1; YSA1H; YSAH1
UniProt ID: (Human) Q9UKK9
Entrez Gene ID: (Human) 11164
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support