Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Reconstitute at 0.5 mg/mL in sterile PBS.
Cytokine that may trigger an antiviral response activating the JAK-STAT pathway and up-regulating specifically some interferon-stimulated genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IFN-lambda-4; IFNAN; IFNL4; IFNL4 splicing form 1 with deletion of exon 2; IFNL4 splicing form 2 with deletion of exons 2 and 3; IFNL4 splicing form 3 with alternative C-terminal domain (exons 4 and 5); IFNL4_HUMAN; Interferon; Interferon lambda-4; interferon, lambda 4; no similarity to known proteins; similar to type III interferons (IFNL1, IFNL2, IFNL3)
Gene Aliases: IFNAN; IFNL4
UniProt ID: (Human) K9M1U5
Entrez Gene ID: (Human) 101180976
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support