Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 12
Positive Control: MCF7 cell lysate, HepG2 cell lysate, SH-SY5Y cell lysate, mouse placenta tissue lysate, rat embryo tissue lysate, human pancreas tissue, mouse brain tissue, rat brain tissue.
Tissue Specificity: Group enriched (adrenal gland, pituitary gland, placenta).
Subcellular Location: Membrane, Cytoplasm.
Sequence Similarities: 83% Mouse/Rat.
Predicted band size: 41 kDa
The Delta-like 1 homolog (DLK1) is a transmembrane protein containing six epidermal growth factor repeats. It is involved in the differentiation of several cell types, including adipocytes and is also thought to be a tumor suppressor. The DLK1 gene is one of several imprinted genes located in a region on chromosome 14q32; certain mutations in this imprinted region can cause phenotypes similar to maternal and paternal uniparental disomy of chromosome 14 (UPD14). DLK1 is expressed from the paternal allele; a polymorphism within this gene has been associated with child and adolescent obesity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adenoma-associated variant of dlk1 protein; Adipocyte differentiation inhibitor protein; alternatively spliced; lacking 219 bp between positions 858 and 859; The 219 bp deletion has been demonstrated to originate by alternative splicing within an exon.; delta-like 1 homolog; dlk; DLK-1; EGF like function; fetal antigen 1; pG2; Preadipocyte factor 1; preadipocyte factor-1; PREF; Pref-1; Protein delta homolog 1; secredeltin; stromal cell derived protein 1; unnamed protein product
Gene Aliases: AW742678; Delta1; DLK; DLK-1; DLK1; DlkI; FA1; Ly107; Peg9; pG2; Pref-1; PREF1; Scp-1; SCP1; ZOG
UniProt ID: (Human) P80370, (Mouse) Q09163, (Rat) O70534
Entrez Gene ID: (Human) 8788, (Mouse) 13386, (Rat) 114587
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support