Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GATOR complex protein SEH1; GATOR2 complex protein SEH1; Nucleoporin SEH1; nucleoporin SEH1-like; Nup107-160 subcomplex subunit SEH1; sec13 like protein; SEC13-like protein; unnamed protein product
Gene Aliases: 2610007A16Rik; AW540070; SEC13L; SEH1; SEH1A; SEH1B; SEH1L
UniProt ID: (Human) Q96EE3, (Mouse) Q8R2U0
Entrez Gene ID: (Rat) 100911798, (Human) 81929, (Mouse) 72124
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support