Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Enolase 1 is a multifunctional enzyme that, as well as its role in glycolysis, plays a part in various processes such as growth control, hypoxia tolerance and allergic responses. May also function in the intravascular and pericellular fibrinolytic system due to its ability to serve as a receptor and activator of plasminogen on the cell surface of several cell-types such as leukocytes and neurons. Stimulates immunoglobulin production.MBP1 binds to the myc promoter and acts as a transcriptional repressor. May be a tumor suppressor.Enolase 2 has neurotrophic and neuroprotective properties on a broad spectrum of central nervous system (CNS) neurons. Binds, in a calcium-dependent manner, to cultured neocortical neurons and promotes cell survival.Enolase 3 appears to have a function in striated muscle development and regeneration.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-phospho-D-glycerate hydro-lyase; 2-phospho-D-glycerate hydrolase; 2-phospho-D-glycerate hydrolyase; alpha enolase; alpha enolase (EC 4.2.1.11); alpha enolase like 1; Alpha-enolase; Beta-enolase; c-Myc promoter binding protein; C-myc promoter-binding protein; ENO1 intronic transcript 1; ENO1 intronic transcript 1 (non-protein coding); ENO2; Enolase 1; enolase 1, (alpha); enolase 1, alpha non-neuron; Enolase 2; enolase 2 (gamma, neuronal); Enolase 3; enolase 3 (beta, muscle); enolase-alpha; epididymis secretory protein Li 17; epididymis secretory protein Li 279; gamma enolase; Gamma-enolase; MBP-1; MPB-1; MSE; muscle enriched enolase; Muscle specific enolase (beta beta enolose) (two splicing products); Muscle-specific enolase; MYC promoter-binding protein 1; Neural enolase; neuron specific gamma enolase; Neuron-specific enolase; neuronal enriched enolase; neurone-specific enolase; NNE; Non-neural enolase; NSE; Phosphopyruvate hydratase; Plasminogen-binding protein; Skeletal muscle enolase; tau-crystallin; transcription factor; unnamed protein product
Gene Aliases: 0610008I15; AI837106; AL022784; BBE; D6Ertd375e; Eno-1; Eno-2; Eno-3; ENO1; ENO1-IT1; ENO1L1; ENO2; ENO3; GSD13; HEL-S-17; HEL-S-279; MBP-1; MBPB1; MPB1; MSE; NNE; NSE; PPH; RNEN3
UniProt ID: (Human) P06733, (Human) P09104, (Human) P13929, (Mouse) P17182, (Rat) P04764, (Mouse) P17183, (Rat) P07323, (Mouse) P21550, (Rat) P15429
Entrez Gene ID: (Human) 2023, (Human) 2026, (Human) 2027, (Mouse) 13806, (Rat) 24333, (Mouse) 13807, (Rat) 24334, (Mouse) 13808, (Rat) 25438
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support