Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LYPVGYLDKQ VPDT
May mediate anti-apoptotic activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell proliferation-inducing gene 17 protein; ER membrane protein complex subunit 4; PIG17; Transmembrane protein 85; unnamed protein product
Gene Aliases: 2610318K02Rik; EMC4; HSPC184; PIG17; RGD1310905; TMEM85
UniProt ID: (Human) Q5J8M3, (Mouse) Q9CZX9
Entrez Gene ID: (Human) 51234, (Rat) 296049, (Chinese hamster) 100772555, (Mouse) 68032
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support