Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • CCL5 (RANTES) Antibodies

          Invitrogen

          CCL5 (RANTES) Polyclonal Antibody

          Advanced Verification
          9 Published Figures
          8 References
          View all (31) CCL5 (RANTES) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CCL5 (RANTES) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Published Figures (9)
          • Advanced Verification (1)
          CCL5 (RANTES) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CCL5 (RANTES) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CCL5 (RANTES) Antibody (P230E) in ICC/IF

          Journal of neuroinflammation 2013 - Figure 2 Immunocytochemistry for HIV-1 Vpr-mediated induction of CCL5 in astrocytes. SVGA astrocytes were cultured on cover slips and were either mock transfected (D-F) or were transfected with a plasmid encoding Vpr (G-I) . Non-transfected cells were used as control (A-C) . The cells were stained for nucleus ( blue ); CCL5 ( green ) and GFAP ( red ), and the images were captured using a Leica TCS SP5 II on... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CCL5 (RANTES) Antibody in Immunocytochemistry (ICC/IF)
          CCL5 (RANTES) Antibody in Immunocytochemistry (ICC/IF)
          CCL5 (RANTES) Antibody in Immunohistochemistry (IHC)
          CCL5 (RANTES) Antibody in Western Blot (WB)
          CCL5 (RANTES) Antibody in ELISA (ELISA)
          CCL5 (RANTES) Antibody in ELISA (ELISA)
          CCL5 (RANTES) Antibody in ELISA (ELISA)
          CCL5 (RANTES) Antibody in ELISA (ELISA)
          CCL5 (RANTES) Antibody in ELISA (ELISA)
          CCL5 (RANTES) Antibody

          Product Details

          P230E

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 2 publications 2 publications

          ELISA (ELISA)

          Assay-dependent
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human

          Published species

          Cynomolgus monkey, Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant human RANTES

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.0 mg/mL

          Amount

          500 µg

          Purification

          Protein G

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_223462

          Product Specific Information

          P230E targets RANTES in ELISA, and WB applications and shows reactivity with Human samples.



          The P230E immunogen is recombinant human RANTES.



          P230E detects RANTES which has a predicted molecular weight of approximately 8 kDa.



          The P230E Rantes polyclonal antibody has successfully been paired as the coating antibody in a sandwich ELISA with detection antibody M230B (biotinylated conjugate of clone VL-1) Typical dilutions for sandwich ELISA include 1 µg/mL for coating and 0.125 - 0.25 µg/mL for detection.



          Antibody P230E and biotinylated antibody M230B (clone VL-1) have successfully been used in combination with recombinant Rantes protein SRNTS in ELISA applications.

          Target Information

          CCL5 (RANTES) is a chemokine gene located on the q-arm of chromosome 17. Chemokines are a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of the N-terminal cysteine residues of the mature peptide. CCL5, a member of the CC subfamily, functions as a chemoattractant for blood monocytes, memory T helper cells, and eosinophils. It causes the release of histamine from basophils and activates eosinophils. This cytokine is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor chemokine (C-C motif) receptor 5 (CCR5), and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor. Alternative splicing results in multiple transcript variants that encode different isoforms. Diseases associated with CCL5 include Human Immunodeficiency Virus Type 1 and Periapical Granuloma.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: beta-chemokine RANTES; C-C motif chemokine 5; chemokine (C-C motif) ligand 5; EoCP; Eosinophil chemotactic cytokine; MGC17164; OTTHUMP00000197106; regulated upon activation, normally T-expressed, and presumably secreted; SIS δ; SIS-delta; SISδ; small inducible cytokine subfamily A (Cys-Cys), member 5; Small-inducible cytokine A5; T cell-specific protein P228; T cell-specific protein precursor; T-cell specific protein p288; T-cell-specific protein RANTES; TCP228; unnamed protein product

          View more View less

          Gene Aliases: CCL5; D17S136E; eoCP; RANTES; SCYA5; SIS-delta; SISd; TCP228

          View more View less

          UniProt ID: (Human) P13501

          View more View less

          Entrez Gene ID: (Human) 6352

          View more View less

          Function(s)
          phosphatidylinositol phospholipase C activity protein kinase activity cytokine activity protein binding chemokine activity phospholipase activator activity receptor signaling protein tyrosine kinase activator activity CCR1 chemokine receptor binding CCR4 chemokine receptor binding CCR5 chemokine receptor binding chemoattractant activity chemokine receptor binding identical protein binding protein homodimerization activity chemokine receptor antagonist activity CCR chemokine receptor binding
          Process(es)
          dendritic cell chemotaxis monocyte chemotaxis regulation of chronic inflammatory response calcium ion transport cellular calcium ion homeostasis exocytosis chemotaxis inflammatory response immune response leukocyte cell-cell adhesion G-protein coupled receptor signaling pathway cell-cell signaling response to virus response to toxic substance positive regulation of macrophage chemotaxis positive regulation of T cell chemotaxis positive regulation of smooth muscle cell migration positive regulation of cell migration phospholipase D-activating G-protein coupled receptor signaling pathway positive regulation of TOR signaling positive regulation of cell-cell adhesion mediated by integrin response to cytokine positive regulation of homotypic cell-cell adhesion response to tumor necrosis factor chemokine (C-C motif) ligand 5 signaling pathway positive regulation of T cell proliferation neutrophil activation negative regulation by host of viral transcription cellular response to fibroblast growth factor stimulus positive regulation of viral genome replication negative regulation of viral genome replication positive regulation of innate immune response negative regulation of G-protein coupled receptor protein signaling pathway positive regulation of G-protein coupled receptor protein signaling pathway positive regulation of cell adhesion positive regulation of translational initiation eosinophil chemotaxis macrophage chemotaxis positive regulation of smooth muscle cell proliferation epithelial cell proliferation positive regulation of epithelial cell proliferation regulation of insulin secretion regulation of T cell activation positive chemotaxis positive regulation of protein kinase B signaling positive regulation of calcium ion transport antimicrobial humoral immune response mediated by antimicrobial peptide chemokine-mediated signaling pathway negative regulation of chemokine-mediated signaling pathway negative regulation of T cell apoptotic process positive regulation of T cell apoptotic process ERK1 and ERK2 cascade cellular response to interferon-gamma cellular response to interleukin-1 cellular response to tumor necrosis factor positive regulation of monocyte chemotaxis receptor signaling pathway via STAT cellular response to virus positive regulation of neuroinflammatory response positive regulation of STAT cascade negative regulation of macrophage apoptotic process positive regulation of T cell migration positive regulation of natural killer cell chemotaxis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline