Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • iNOS Antibodies

          Invitrogen

          iNOS Polyclonal Antibody

          Advanced Verification
          68 Published Figures
          189 References
          View all (53) iNOS antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite iNOS Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          • Published Figures (68)
          • Advanced Verification (1)
          iNOS Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          iNOS Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 75

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          iNOS Antibody (PA1-036) in ICC/IF

          Immunofluorescent analysis of iNOS in NIH-3T3 Cells. Cells were grown on chamber slides and fixed with formaldehyde prior to staining. Cells were probed without (control) or with a iNOS polyclonal antibody (Product # PA1-036) at a dilution of 1:20 overnight at 4 C, washed with PBS and incubated with a DyLight-488 conjugated secondary antibody (Product # 35552). iNOS staining (green), F-Actin staining with Phalloidin (red) and nuclei with DAPI (blue) is shown. Images were taken at 60X magnification. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          iNOS Antibody in Immunocytochemistry (ICC/IF)
          iNOS Antibody in Immunocytochemistry (ICC/IF)
          iNOS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          iNOS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Immunohistochemistry (Paraffin), Immunocytochemistry (IHC (P), ICC/IF)
          iNOS Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          iNOS Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          iNOS Antibody in Immunocytochemistry (ICC/IF)
          iNOS Antibody in Immunocytochemistry (ICC/IF)
          iNOS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          iNOS Antibody in Immunohistochemistry (IHC)
          iNOS Antibody in Immunohistochemistry (IHC)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Immunocytochemistry (ICC/IF)
          iNOS Antibody in Immunocytochemistry (ICC/IF)
          iNOS Antibody in Immunocytochemistry (ICC/IF)
          iNOS Antibody in Immunocytochemistry (ICC/IF)
          iNOS Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          iNOS Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          iNOS Antibody in Immunohistochemistry (Frozen) (IHC (F))
          iNOS Antibody in Immunohistochemistry (Frozen) (IHC (F))
          iNOS Antibody in Immunohistochemistry (IHC)
          iNOS Antibody in Immunohistochemistry (IHC)
          iNOS Antibody in Immunohistochemistry (IHC)
          iNOS Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          iNOS Antibody in Immunohistochemistry (IHC)
          iNOS Antibody in Immunohistochemistry (IHC)
          iNOS Antibody in Immunohistochemistry (IHC)
          iNOS Antibody in Immunohistochemistry (IHC)
          iNOS Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          iNOS Antibody in Immunohistochemistry (IHC)
          iNOS Antibody in Immunohistochemistry (IHC)
          iNOS Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot, Flow Cytometry (WB, Flow)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Western Blot (WB)
          iNOS Antibody in Flow Cytometry (Flow)
          iNOS Antibody in Flow Cytometry (Flow)
          iNOS Antibody in Immunomicroscopy (IM)
          iNOS Antibody
          iNOS Polyclonal Antibody

          Product Details

          PA1-036

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200-1:1,000
          View 77 publications 77 publications

          Immunohistochemistry (IHC)

          -
          View 81 publications 81 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20
          View 7 publications 7 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 3 publications 3 publications

          Immunocytochemistry (ICC/IF)

          1:50
          View 18 publications 18 publications

          Flow Cytometry (Flow)

          -
          View 1 publication 1 publication

          Immunomicroscopy (IM)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Amphibian, Avian, Cat, Fish, Human, Mouse, Pig, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues D(17) L K E E K D I N N N V K K T(31) of mouse iNOS.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          4 mg/mL

          Amount

          800 µg

          Purification

          Ammonium sulfate precipitation

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_325773

          Product Specific Information

          PA1-036 detects inducible nitric oxide synthase (iNOS) from human, mouse and rat tissues and cells as well as recombinant human and mouse iNOS. This antibody does not detect other NOS isoforms.

          PA1-036 has been successfully used in Western blot and immunofluorescence procedures. By Western blot, this antibody detects an ~135 kDa protein representing mouse iNOS from LPS stimulated RAW 264.7 cells.

          The PA1-036 immunogen is a synthetic peptide corresponding to residues D(17) L K E E K D I N N N V K K T(31) of mouse iNOS.

          Target Information

          iNOS (Inducible Nitric oxide, NO, NOS) is an inorganic, gaseous free radical that carries a variety of messages between cells. Vasorelaxation, neurotransmission and cytotoxicity can all be potentiated through cellular response to NO. NO production is mediated by members of the nitric oxide synthase (NOS) family. iNOS is expressed in liver and inducible by a combination of lipopolysaccharide and certain cytokines. NOS catalyzes the oxidization of L-arginine to produce L-citrulline and NO. Two constitutive isoforms, brain or neuronal NOS (b or nNOS, type I) and endothelial cell NOS (eNOS, type III), and one inducible isoform (iNOS, type II), have been cloned. All NOS isoforms contain calmodulin, nicotinamide adenine dinucleotide phosphate (NADPH), flavin adenine dinucleotide (FAD), and flavin mononucleotide (FMN) binding domains. iNOS is found in a variety of cell types including macrophages, hepatocytes, synoviocytes, and smooth muscle cells. Cytokines such as interferon-gamma (IFN), tumor necrosis factor (TNF), interleukin-1 and -2, and lipopolysaccarides (LPS) cause an increase in iNOS mRNA, protein, and activity levels. Protein kinase C-stimulating agents exhibit the same effect on iNOS activity. After cytokine induction, iNOS exhibits a delayed activity response which is then followed by a significant increase in NO production over a long period of time. Three related iNOS pseudogenes are located within the Smith-Magenis syndrome region on chromosome 17. Diseases associated with iNOS dysfunction include achalasia and impotence.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: calcium-calmodulin independent nitric oxide synthase iNOS; HEP-NOS; Hepatocyte NOS; hepatocytes; inducible nitric oxide synthase; Inducible NO synthase; Inducible NOS; MAC-NOS; Macrophage NOS; nitric oxide synthase; nitric oxide synthase 2, inducible; nitric oxide synthase 2, inducible, macrophage; nitric oxide synthase 2A (inducible, hepatocytes); Nitric oxide synthase, inducible; nitric oxide synthase, macrophage; nitric oxide synthase-inducible; NOS type II; NOS, type II; OTTMUSP00000000202; Peptidyl-cysteine S-nitrosylase NOS2; unnamed protein product

          View more View less

          Gene Aliases: HEP-NOS; i-NOS; INOS; Inosl; NOS; Nos-2; NOS-II; NOS2; NOS2A

          View more View less

          UniProt ID: (Human) P35228, (Mouse) P29477, (Rat) Q06518

          View more View less

          Entrez Gene ID: (Human) 4843, (Mouse) 18126, (Rat) 24599

          View more View less

          Function(s)
          nitric-oxide synthase activity protein binding calmodulin binding FMN binding oxidoreductase activity heme binding tetrahydrobiopterin binding arginine binding protein homodimerization activity metal ion binding flavin adenine dinucleotide binding NADP binding actin binding NADPH-hemoprotein reductase activity receptor binding iron ion binding beta-catenin binding cAMP-dependent protein kinase regulator activity protein kinase binding cadherin binding nitric-oxide synthase binding Hsp90 protein binding amino acid binding oxidoreductase protein modifying enzyme
          Process(es)
          response to hypoxia positive regulation of leukocyte mediated cytotoxicity innate immune response in mucosa arginine catabolic process superoxide metabolic process nitric oxide biosynthetic process inflammatory response nitric oxide mediated signal transduction circadian rhythm response to bacterium response to hormone negative regulation of gene expression peptidyl-cysteine S-nitrosylation prostaglandin secretion response to lipopolysaccharide positive regulation of interleukin-6 production positive regulation of interleukin-8 production Fc-gamma receptor signaling pathway involved in phagocytosis regulation of cell proliferation negative regulation of protein catabolic process defense response to bacterium regulation of cellular respiration cell redox homeostasis negative regulation of blood pressure regulation of insulin secretion defense response to Gram-negative bacterium cellular response to lipopolysaccharide cellular response to interferon-gamma cellular response to xenobiotic stimulus regulation of cytokine production involved in inflammatory response ovulation from ovarian follicle positive regulation of guanylate cyclase activity cellular response to drug regulation of protein kinase activity positive regulation of vasodilation positive regulation of killing of cells of other organism oxidation-reduction process endothelial cell proliferation blood vessel remodeling signal transduction G-protein coupled receptor signaling pathway coupled to cGMP nucleotide second messenger aging regulation of heart contraction regulation of blood pressure response to mechanical stimulus response to activity glycoside metabolic process cGMP-mediated signaling response to estradiol response to tumor necrosis factor intracellular signal transduction response to immobilization stress wound healing positive regulation of apoptotic process retinoic acid receptor signaling pathway cellular response to lithium ion cellular response to vitamin D cellular response to interleukin-1 cellular response to tumor necrosis factor cellular response to redox state response to dexamethasone
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline