Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Caveolin 3 Antibodies

          Invitrogen

          Caveolin 3 Polyclonal Antibody

          Advanced Verification
          10 Published Figures
          9 References
          View all (16) Caveolin 3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Caveolin 3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (8)
          • Published Figures (10)
          • Advanced Verification (1)
          Caveolin 3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Caveolin 3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 19

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Caveolin 3 Antibody (PA1-066) in WB

          Western blot analysis was performed on whole cell extracts (20 µg lysate) of Rat Heart (Lane 1), Mouse Heart (Lane 2), HEK-293 (Lane 3), Rat Skeletal Muscle (lane 4) and Mouse Muscle (lane 5). The blots were probed with Anti-Caveolin 3 Rabbit Polyclonal Antibody (Product # PA1-066, 1-3 µg/mL) and detected by chemiluminescence using Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibody, HRP conjugate (Product # A27036, 0.4 µg/mL, 1:2500 dilution). A 17 kDa band corresponding to Caveolin 3 was observed across tissues tested exce... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Caveolin 3 Antibody in Western Blot (WB)
          Caveolin 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Caveolin 3 Antibody in Immunoprecipitation (IP)
          Caveolin 3 Antibody in Immunocytochemistry (ICC/IF)
          Caveolin 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Caveolin 3 Antibody in Immunocytochemistry (ICC/IF)
          Caveolin 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Caveolin 3 Antibody in Western Blot (WB)
          Caveolin 3 Antibody in Immunocytochemistry (ICC/IF)
          Caveolin 3 Antibody in Immunocytochemistry (ICC/IF)
          Caveolin 3 Antibody in Western Blot (WB)
          Caveolin 3 Antibody in Western Blot (WB)
          Caveolin 3 Antibody in Western Blot (WB)
          Caveolin 3 Antibody in Western Blot (WB)
          Caveolin 3 Antibody in Western Blot (WB)
          Caveolin 3 Antibody in Western Blot (WB)
          Caveolin 3 Antibody in Western Blot (WB)
          Caveolin 3 Antibody in Western Blot (WB)
          Caveolin 3 Antibody
          Caveolin 3 Polyclonal Antibody

          Product Details

          PA1-066

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-3 µg/mL
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:200
          -

          Immunocytochemistry (ICC/IF)

          1-2 µg/mL
          View 6 publications 6 publications

          Immunoprecipitation (IP)

          5 µg
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Mouse, Non-human primate, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues M(1) M T E E H T D L E A R I I K D I H C(19) C of mouse CAV3
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2072446

          Product Specific Information

          PA1-066 detects Caveolin 3 from rat and mouse tissues. This antibody does not detect caveolin-1 or -2.

          PA1-066 has been successfully used in Western blot, IF, immunocytochemistry and immunoprecipitation procedures. By Western blot, this antibody detects an ~21 kDa protein representing Caveolin 3 from rat heart homogenate.

          The PA1-066 immunogen is a synthetic peptide corresponding to residues M(1) M T E E H T D L E A R I I K D I H C(19) C of mouse CAV3. This sequence is ~85% conserved in human Caveolin 3. PA1-066 immunizing peptide (Cat. # PEP-081) is available for use in neutralization and control experiments.

          Target Information

          Caveolin-3 (CAV3) acts as a scaffolding protein within the caveolar membrane. It interacts directly with G-protein alpha subunits and can functionally regulate their activity. Caveolin-3 may also regulate voltage-gated potassium channels. It also plays a role in the sarcolemma repair mechanism of both skeletal muscle and cardiomyocytes that permits rapid resealing of membranes disrupted by mechanical stress. Caveolin-3 also mediates the recruitment of CAVIN2 and CAVIN3 proteins to the caveolae. Mutations in the gene can result in HyperCKmia, rippling muscle disease 2, cardiomyopathy, long QT syndrome 9, sudden infant death syndrome, and myopathy.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CAV; Caveolin-3; cavolin 3; contains K15N mutation; M-caveolin; MGC126100; MGC126101; MGC126129; unnamed protein product

          View more View less

          Gene Aliases: AI385751; Cav-3; CAV3; LGMD1C; LQT9; M-cav; MPDT; RMD2; VIP-21; VIP21

          View more View less

          UniProt ID: (Human) P56539, (Rat) P51638, (Mouse) P51637

          View more View less

          Entrez Gene ID: (Human) 859, (Rat) 29161, (Mouse) 12391

          View more View less

          Function(s)
          calcium channel regulator activity protein binding sodium channel regulator activity potassium channel inhibitor activity enzyme binding alpha-tubulin binding ion channel binding macromolecular complex binding nitric-oxide synthase binding binding, bridging connexin binding protein C-terminus binding protein complex binding protein complex scaffold scaffold/adaptor protein
          Process(es)
          MAPK cascade response to hypoxia plasma membrane repair regulation of heart rate response to ischemia cardiac muscle hypertrophy triglyceride metabolic process calcium ion transport endocytosis plasma membrane organization actin filament organization positive regulation of cytosolic calcium ion concentration muscle organ development myoblast fusion regulation of heart contraction protein localization positive regulation of cell proliferation negative regulation of cardiac muscle hypertrophy positive regulation of myotube differentiation regulation of skeletal muscle contraction myotube differentiation regulation of transforming growth factor beta receptor signaling pathway cell differentiation positive regulation of microtubule polymerization cytoplasmic microtubule organization membrane raft organization T-tubule organization detection of muscle stretch regulation of signal transduction by receptor internalization regulation of membrane potential glucose homeostasis cholesterol homeostasis negative regulation of MAPK cascade negative regulation of cell size regulation of nerve growth factor receptor activity regulation of cytosolic calcium ion concentration nucleus localization establishment of localization in cell regulation of protein kinase B signaling regulation of calcium ion transport negative regulation of calcium ion transport cardiac muscle cell development regulation of cardiac muscle contraction negative regulation of sarcomere organization regulation of ventricular cardiac muscle cell membrane repolarization heart trabecula formation regulation of ventricular cardiac muscle cell membrane depolarization regulation of branching involved in mammary gland duct morphogenesis negative regulation of cell growth involved in cardiac muscle cell development caveola assembly protein localization to plasma membrane ventricular cardiac muscle cell action potential regulation of calcium ion import regulation of cardiac muscle cell action potential involved in regulation of contraction regulation of p38MAPK cascade regulation of membrane depolarization during cardiac muscle cell action potential negative regulation of membrane depolarization during cardiac muscle cell action potential negative regulation of potassium ion transmembrane transport regulation of sodium ion transmembrane transport cellular response to ionomycin negative regulation of protein localization to cell surface positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process positive regulation of caveolin-mediated endocytosis negative regulation of protein kinase activity cell growth negative regulation of MAP kinase activity muscle cell cellular homeostasis negative regulation of nitric-oxide synthase activity establishment of protein localization to plasma membrane negative regulation of potassium ion transmembrane transporter activity regulation of calcium ion transmembrane transporter activity regulation of sodium ion transmembrane transporter activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline