Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • KDEL Antibodies

          Invitrogen

          KDEL Polyclonal Antibody

          Advanced Verification
          12 Published Figures
          27 References
          View all (23) KDEL antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite KDEL Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          • Published Figures (12)
          • Advanced Verification (1)
          KDEL Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          KDEL Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 18

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          KDEL Antibody (PA1-013) in ICC/IF

          Immunofluorescent analysis of KDEL in U251 Cells. Cells were grown on chamber slides and fixed with formaldehyde prior to staining. Cells were probed without (control) or with a KDEL polyclonal antibody (Product # PA1-013) at a dilution of 1:200 overnight at 4 C, washed with PBS and incubated with a DyLight-488 conjugated secondary antibody (Product # 35552). KDEL staining (green), F-Actin staining with Phalloidin (red) and nuclei with DAPI (blue) is shown. Images were taken at 60X magnification. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          KDEL Antibody in Immunocytochemistry (ICC/IF)
          KDEL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KDEL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KDEL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KDEL Antibody in Flow Cytometry (Flow)
          KDEL Antibody in Immunocytochemistry (ICC/IF)
          KDEL Antibody in Immunocytochemistry (ICC/IF)
          KDEL Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          KDEL Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          KDEL Antibody in Immunohistochemistry (IHC)
          KDEL Antibody in Immunohistochemistry (IHC)
          KDEL Antibody in Immunohistochemistry (IHC)
          KDEL Antibody in Immunohistochemistry (IHC)
          KDEL Antibody in Western Blot (WB)
          KDEL Antibody in Western Blot (WB)
          KDEL Antibody in Western Blot (WB)
          KDEL Antibody in Western Blot (WB)
          KDEL Antibody
          KDEL Polyclonal Antibody

          Product Details

          PA1-013

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          8-16 µg/mL
          View 4 publications 4 publications

          Immunohistochemistry (IHC)

          -
          View 6 publications 6 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:200
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 15 publications 15 publications

          Flow Cytometry (Flow)

          1-20 µg/mL
          -

          Immunoprecipitation (IP)

          Assay-dependent
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Hamster, Human, Mouse, Rat

          Published species

          Hamster, Human, Mouse, Non-human primate, Rat, Sea urchin

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues C T(643) G E E D T S E K D E L(654) of rat GRP94.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_325593

          Product Specific Information

          PA1-013 detects KDEL from human, rat, mouse and hamster.

          PA1-013 has been successfully used in Western blot, immunofluorescence and immunoprecipitation procedures. By Western blot, this antibody detects a 57 kDa protein representing protein disulfide isomerase (PDI), a 78 kDa protein representing glucose regulated protein 70 kDa (GRP78) and an ~94 kDa protein representing GRP94 in mouse myeloma cells. Immunofluorescence staining of GRP78 in mouse myeloma cells with PA1-013 results in diffuse cytoplasmic staining consistent with specific ER localization. PA1-013 immunoprecipitates PDI, GRP78, an ~72 kDa protein which may be endoplasmic reticulum protein 72 kDa (ERp72), and an ~94 kDa protein representing GRP94 from numerous rodent tissues.

          The PA1-013 immunogen is a synthetic peptide corresponding to residues C T(643) G E E D T S E K D E L(654) of rat GRP94.

          Reconstitute in 100 µL PBS to create a stock of 1 mg/mL.

          Target Information

          The sequence Lys-Asp-Glu-Leu (KDEL) or a closely related sequence, is present at the carboxy-terminus of soluble endoplasmic reticulum (ER) resident proteins and some membrane proteins. 78 and 94 kDa glucose regulated proteins, GRP78 and GRP94 respectively, and protein disulfide isomerase (PDI) all share the C-terminal KDEL sequence. The presence of carboxy-terminal KDEL appears to be necessary for ER retention and appears to be sufficient to reduce the secretion of proteins from the ER. This retention is reported to be mediated by a KDEL receptor.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 78 kDa glucose-regulated protein; 94 kDa glucose-regulated protein; 94 kDa glucose-regulated protein {ECO:0000250|UniProtKB:P08113}; Binding-immunoglobulin protein; BiP; chaperone; Endoplasmic reticulum chaperone BiP; endoplasmic reticulum lumenal Ca(2+)-binding protein grp78; Endoplasmic reticulum resident protein 99; Endoplasmin; endoplasmin {ECO:0000250|UniProtKB:P08113}; endothelial cell (HBMEC) glycoprotein; epididymis luminal protein 35; epididymis secretory sperm binding protein Li 125m; epididymis secretory sperm binding protein Li 89n; ER lumen protein retaining receptor 1; ER lumen protein-retaining receptor 1; ERp99; glucose regulated protein, 78 kDa; glucose-regulated protein; glucose-regulated protein (grp78); glucose-regulated protein, 78kDa; gp96 homolog; GRP 78; GRP-78; GRP-94; heat shock 70 kDa protein 5; heat shock 70kD protein 5; heat shock 70kD protein 5 (glucose-regulated protein, 78kD); heat shock 70kDa protein 5 (glucose-regulated protein); heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa); heat shock protein 5; Heat shock protein 70 family protein 5; Heat shock protein 90 kDa beta member 1; heat shock protein 90 kDa beta member 1 {ECO:0000250|UniProtKB:P08113}; heat shock protein 90, beta (Grp94), member 1; heat shock protein 90kDa beta (Grp94), member 1; heat shock protein 90kDa beta family member 1; Heat shock protein family A member 5; Heat shock protein family C member 4; HSP70 family protein 5; hsp90b1 {ECO:0000250|UniProtKB:P08113}; Immunoglobulin heavy chain-binding protein; KDEL; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1; KDEL endoplasmic reticulum protein retention receptor 1; KDEL receptor 1; Lys-Asp-Glu-Leu; Polymorphic tumor rejection antigen 1; Putative MAPK-activating protein PM23; RP23-446N16.1; Steroidogenesis-activator polypeptide; stress-inducible tumor rejection antigen gp96; transforming growth factor alpha regulated gene 2; tumor rejection antigen (gp96) 1; Tumor rejection antigen 1; Tumor rejection antigen gp96; unnamed protein product; XAP-1 antigen

          View more View less

          Gene Aliases: 8030486F04Rik; AL022860; AU019543; AW215843; baffled; BIP; D2Wsu141e; D2Wsu17e; ECGP; endoplasmin; ERD2; ERD2.1; ERp99; GP96; GRP78; GRP94; HDEL; HEL-S-125m; HEL-S-89n; HEL35; Hsce70; HSP90B1; HSPA5; HSPC4; I79_019946; I79_020931; KDELR1; mBiP; PM23; SEZ-7; Sez7; TA-3; Targ2; Tra-1; TRA1

          View more View less

          UniProt ID: (Human) P14625, (Human) P11021, (Mouse) P08113, (Rat) Q66HD0, (Rat) P06761, (Mouse) P20029, (Chinese hamster) G3I8R9, (Human) P24390, (Mouse) Q99JH8, (Rat) Q569A6

          View more View less

          Entrez Gene ID: (Human) 7184, (Human) 3309, (Chinese hamster) 100773331, (Mouse) 22027, (Rat) 362862, (Rat) 25617, (Mouse) 14828, (Chinese hamster) 100689305, (Human) 10945, (Mouse) 68137, (Rat) 361577, (Chinese hamster) 100750912

          View more View less

          Function(s)
          nucleotide binding RNA binding protein phosphatase inhibitor activity calcium ion binding protein binding ATP binding hydrolase activity ATPase activity protein phosphatase binding protein binding involved in protein folding low-density lipoprotein particle receptor binding unfolded protein binding ATP-dependent protein folding chaperone enzyme binding protein domain specific binding protein serine/threonine kinase inhibitor activity heat shock protein binding ubiquitin protein ligase binding ribosome binding cadherin binding chaperone binding misfolded protein binding virion binding glycoprotein binding KDEL sequence binding ER retention sequence binding molecular_function Hsp90 family chaperone membrane trafficking regulatory protein
          Process(es)
          response to hypoxia protein folding protein transport positive regulation of Wnt signaling pathway retrograde protein transport, ER to cytosol actin rod assembly positive regulation of toll-like receptor signaling pathway protein folding in endoplasmic reticulum response to endoplasmic reticulum stress ERAD pathway negative regulation of apoptotic process sequestering of calcium ion cellular response to manganese ion cellular response to ATP protein localization to plasma membrane translational initiation ER overload response cerebellum structural organization cerebellar Purkinje cell layer development substantia nigra development positive regulation of cell migration negative regulation of transforming growth factor beta receptor signaling pathway endoplasmic reticulum unfolded protein response posttranslational protein targeting to membrane, translocation negative regulation of protein complex assembly positive regulation of protein ubiquitination maintenance of protein localization in endoplasmic reticulum IRE1-mediated unfolded protein response PERK-mediated unfolded protein response protein refolding cellular response to glucose starvation positive regulation of transcription from RNA polymerase II promoter proteolysis involved in cellular protein catabolic process regulation of protein folding in endoplasmic reticulum cellular response to interleukin-4 regulation of ATF6-mediated unfolded protein response regulation of IRE1-mediated unfolded protein response negative regulation of IRE1-mediated unfolded protein response regulation of PERK-mediated unfolded protein response negative regulation of PERK-mediated unfolded protein response response to stress ER-associated ubiquitin-dependent protein catabolic process regulation of phosphoprotein phosphatase activity activation of signaling protein activity involved in unfolded protein response cellular response to antibiotic toxin transport positive regulation of embryonic development protein retention in ER lumen retrograde vesicle-mediated transport, Golgi to ER vesicle-mediated transport T cell differentiation T cell apoptotic process transport biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline