Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
PA1-028 detects cyclophilin D (CyPD) in rat, hamster, canine, mouse, and human tissues.
PA1-028 has been sucessfully used in Western blot procedures. By Western blot, this antibody detects a ~15 kDa protein in rat liver and heart extract, and a ~37 kDa in rat brain extract.
The PA1-028 immunogen is a synthetic peptide corresponding to residues S(42) G N P L V Y L D V D A N(54) C of mature human CyPD. This peptide (Cat. # PEP-104) is available for use in neutralization and control experiments.
This gene encodes a H1 histone binding protein that is involved in transporting histones into the nucleus of dividing cells. Multiple isoforms are encoded by transcript variants of this gene. The somatic form is expressed in all mitotic cells, is localized to the nucleus, and is coupled to the cell cycle. The testicular form is expressed in embryonic tissues, tumor cells, and the testis. In male germ cells, this protein is localized to the cytoplasm of primary spermatocytes, the nucleus of spermatids, and the periacrosomal region of mature spermatozoa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 40 kDa peptidyl-prolyl cis-trans isomerase; 40 kDa peptidyl-prolyl cis-trans isomerase D; cyclophilin 40; Cyclophilin D; Cyclophilin-40; Cyclophilin-related protein; Cyp 40; Cyp D; CYP-40; CYP-S1; cytoplasmic cyclophilin D; EC 5.2.1.8; MGC33096; Peptidyl-prolyl cis-trans isomerase D; peptidylprolyl isomerase D; peptidylprolyl isomerase D (cyclophilin D); PPIase; PPIase D; PPID; rotamase; Rotamase D; S-cyclophilin; SCYLP; testicular tissue protein Li 147; unnamed protein product
Gene Aliases: 4930564J03Rik; CYP-40; CYP40; CYPD; I79_011990; PPID; Ppidl; Ppif
UniProt ID: (Human) Q08752, (Mouse) Q9CR16, (Rat) Q6DGG0
Entrez Gene ID: (Dog) 475481, (Human) 5481, (Mouse) 67738, (Chinese hamster) 100752921, (Rat) 361967
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support