Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • uNOS Antibodies

          Invitrogen

          uNOS Polyclonal Antibody

          7 Published Figures
          35 References
          View all (3) uNOS antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite uNOS Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (7)
          uNOS Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          uNOS Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          uNOS Antibody (PA1-039) in IHC

          Immunolocalization staining of NOS in drosophila optic nerve using Product # PA1-039. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          uNOS Antibody in Immunohistochemistry (IHC)
          uNOS Antibody in Immunocytochemistry (ICC/IF)
          uNOS Antibody in Immunocytochemistry (ICC/IF)
          uNOS Antibody in Immunohistochemistry (IHC)
          uNOS Antibody in Immunohistochemistry (IHC)
          uNOS Antibody in Immunohistochemistry (IHC)
          uNOS Antibody in Immunohistochemistry (IHC)
          uNOS Antibody in Western Blot (WB)
          uNOS Polyclonal Antibody

          Product Details

          PA1-039

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500
          View 8 publications 8 publications

          Immunohistochemistry (IHC)

          -
          View 16 publications 16 publications

          Immunohistochemistry (Frozen) (IHC (F))

          1:100
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:100-1:200
          View 10 publications 10 publications
          Product Specifications

          Species Reactivity

          Arthropod, Bovine, Fruit fly, Human, Mouse, Pig, Rat

          Published species

          Arthropod, Cnidarian, Crab, Fish, Fruit fly, Human, Insect, Mollusc, Mouse, Pig, Rabbit, Rat, Shark

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues D Q(1113) K R Y H E D I F G(1122) of murine iNOS and bNOS.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_325776

          Product Specific Information

          PA1-039 detects mouse macrophage inducible nitric oxide synthase (iNOS); rat, bovine, Drosophila, and porcine brain nitric oxide synthase (bNOS); human, porcine and bovine epithelial nitric oxide synthase (eNOS).

          PA1-039 has been successfully used in Western blot and immunohistochemistry procedures. By Western blot, this antibody detects an ~135 kDa protein representing mouse macrophage iNOS; an ~155 kDa protein representing rat, bovine and porcine bNOS; ~140 kDa protein representing porcine and bovine eNOS. Immunohistochemical staining of NOS in Drosophila optic tissue with PA1-039 results in staining of the optic nerve. In immunohistochemistry procedures PA1-039 also stains NOS in human endothelial cells, macrophages, and peripheral neural tract cells.

          The PA1-039 immunogen is a synthetic peptide corresponding to residues D Q(1113) K R Y H E D I F G(1122) of mouse iNOS and bNOS. This sequence is highly conserved among the different NOS isoforms.

          Target Information

          NOS oxidizes a guanidine nitrogen of arginine releasing nitric oxide in the form of a free radical and citrulline. Nitric oxide thus generated acts as a messenger in diverse functions including vasodilation neurotransmission, anti-tumor and anti-pathogenic activities. NOS is classified under three types: neuronal NOS (nNOS) or brain NOS (bNOS); induciblr NOS (iNOS) or macrophage NOS (mNOS); and endothelial NOS (eNOS). This antibody reacts with iNOS, bNOS and eNOS.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: bNOS; calcium-calmodulin independent nitric oxide synthase iNOS; cNOS; Constitutive NOS; EC-NOS; endothelial nitric oxide synthase; endothelial nitric oxide synthase 3; Endothelial NOS; eNOS; HEP-NOS; Hepatocyte NOS; inducible nitric oxide synthase; Inducible NO synthase; Inducible NOS; MAC-NOS; Macrophage NOS; N-NOS; NC-NOS; Neuronal NOS; nitric oxidase synthase; nitric oxide synthase; Nitric oxide synthase 1; nitric oxide synthase 1 (neuronal); nitric oxide synthase 2, inducible; nitric oxide synthase 2, inducible, macrophage; nitric oxide synthase 2A (inducible, hepatocytes); Nitric oxide synthase 3; nitric oxide synthase 3 (endothelial cell); Nitric oxide synthase, brain; Nitric oxide synthase, endothelial; Nitric oxide synthase, inducible; nitric oxide synthase, macrophage; nitric-oxide synthase; nNOS; NOS type I; NOS type II; NOS type III; NOS, type II; NOSII; NOSIII; Peptidyl-cysteine S-nitrosylase NOS1; Peptidyl-cysteine S-nitrosylase NOS2; unnamed protein product

          View more View less

          Gene Aliases: 2310005C01Rik; 2310065A03Rik; Bnos; cNOS; EC-NOS; ECNOS; eNOS; HEP-NOS; i-NOS; IHPS1; INOS; Inosl; N-NOS; NC-NOS; nNOS; NO; NOS; Nos-1; Nos-2; Nos-3; NOS-I; NOS-II; NOS1; NOS2; NOS2A; NOS3; NOSIII

          View more View less

          UniProt ID: (Human) P29475, (Human) P35228, (Pig) P79290, (Bovine) Q27995, (Human) P29474, (Pig) Q28969, (Rat) P29476, (Mouse) Q9Z0J4, (Mouse) P29477, (Rat) Q06518, (Mouse) P70313, (Rat) Q62600

          View more View less

          Entrez Gene ID: (Human) 4842, (Pig) 100520934, (Human) 4843, (Pig) 396859, (Bovine) 282876, (Bovine) 287024, (Human) 4846, (Pig) 397557, (Rat) 24598, (Mouse) 18125, (Mouse) 18126, (Rat) 24599, (Mouse) 18127, (Rat) 24600

          View more View less

          Function(s)
          nitric-oxide synthase activity calcium channel regulator activity protein binding calmodulin binding FMN binding oxidoreductase activity sodium channel regulator activity heme binding tetrahydrobiopterin binding arginine binding peptidyl-cysteine S-nitrosylase activity ion channel binding cadmium ion binding metal ion binding calcium-dependent protein binding flavin adenine dinucleotide binding NADP binding scaffold protein binding protein homodimerization activity actin monomer binding superoxide-generating NADPH oxidase activity NADPH-hemoprotein reductase activity iron ion binding amino acid binding enzyme binding ATPase binding actin binding receptor binding beta-catenin binding cAMP-dependent protein kinase regulator activity protein kinase binding cadherin binding nitric-oxide synthase binding Hsp90 protein binding oxidoreductase protein modifying enzyme
          Process(es)
          response to hypoxia regulation of sodium ion transport arginine catabolic process nitric oxide biosynthetic process potassium ion transport calcium ion transport striated muscle contraction cell communication nitric oxide mediated signal transduction myoblast fusion blood circulation response to heat response to hormone negative regulation of calcium ion transport into cytosol regulation of cardiac muscle contraction by calcium ion signaling regulation of metal ion transport signaling response to lipopolysaccharide multicellular organismal response to stress xenobiotic catabolic process vasodilation negative regulation of potassium ion transport negative regulation of ion transport positive regulation of neuron apoptotic process cell redox homeostasis negative regulation of blood pressure positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter regulation of neurogenesis negative regulation of serotonin uptake establishment of localization in cell negative regulation of calcium ion transport regulation of cardiac muscle contraction regulation of postsynaptic membrane potential regulation of biological quality cellular response to growth factor stimulus positive regulation of the force of heart contraction synaptic signaling by nitric oxide positive regulation of adenylate cyclase-activating G protein-coupled receptor signaling pathway positive regulation of sodium ion transmembrane transport regulation of calcium ion transmembrane transport via high voltage-gated calcium channel regulation of blood circulation positive regulation of membrane repolarization during ventricular cardiac muscle cell action potential positive regulation of leukocyte mediated cytotoxicity innate immune response in mucosa superoxide metabolic process inflammatory response circadian rhythm response to bacterium negative regulation of gene expression peptidyl-cysteine S-nitrosylation prostaglandin secretion positive regulation of interleukin-6 production positive regulation of interleukin-8 production Fc-gamma receptor signaling pathway involved in phagocytosis regulation of cell proliferation negative regulation of protein catabolic process defense response to bacterium regulation of cellular respiration regulation of insulin secretion defense response to Gram-negative bacterium cellular response to lipopolysaccharide cellular response to interferon-gamma cellular response to xenobiotic stimulus regulation of cytokine production involved in inflammatory response interleukin-6 production interleukin-8 production oxidation-reduction process mitochondrion organization blood coagulation positive regulation of guanylate cyclase activity cellular response to laminar fluid shear stress negative regulation of extrinsic apoptotic signaling pathway via death domain receptors negative regulation of leukocyte cell-cell adhesion angiogenesis ovulation from ovarian follicle in utero embryonic development blood vessel remodeling regulation of the force of heart contraction by chemical signal regulation of systemic arterial blood pressure by endothelin aortic valve morphogenesis pulmonary valve morphogenesis endocardial cushion morphogenesis regulation of blood pressure negative regulation of cell proliferation negative regulation of platelet activation positive regulation of gene expression negative regulation of muscle hyperplasia smooth muscle hyperplasia removal of superoxide radicals lung development regulation of neurological system process lipopolysaccharide-mediated signaling pathway response to fluid shear stress positive regulation of blood vessel endothelial cell migration endothelial cell migration positive regulation of Notch signaling pathway positive regulation of angiogenesis tetrahydrobiopterin metabolic process nitric oxide metabolic process negative regulation of smooth muscle cell proliferation homeostasis of number of cells within a tissue ventricular septum morphogenesis negative regulation of biomineral tissue development regulation of blood vessel diameter positive regulation of blood vessel diameter muscle contraction female pregnancy aging regulation of heart contraction response to organonitrogen compound response to lead ion response to organic cyclic compound response to activity response to nutrient levels negative regulation of peptidyl-serine phosphorylation response to vitamin E positive regulation of histone acetylation response to nicotine exogenous drug catabolic process negative regulation of apoptotic process response to peptide hormone response to estrogen response to ethanol negative regulation of heart contraction negative regulation of vasoconstriction positive regulation of vasodilation negative regulation of insulin secretion behavioral response to cocaine negative regulation of hydrolase activity negative regulation of cytosolic calcium ion concentration regulation of sensory perception of pain cellular response to mechanical stimulus response to nitric oxide cellular response to epinephrine stimulus positive regulation of long-term synaptic potentiation positive regulation of adrenergic receptor signaling pathway involved in heart process positive regulation of neuron death cellular response to drug regulation of protein kinase activity positive regulation of killing of cells of other organism endothelial cell proliferation signal transduction G-protein coupled receptor signaling pathway coupled to cGMP nucleotide second messenger response to mechanical stimulus glycoside metabolic process cGMP-mediated signaling response to estradiol response to tumor necrosis factor intracellular signal transduction response to immobilization stress wound healing positive regulation of apoptotic process retinoic acid receptor signaling pathway cellular response to lithium ion cellular response to vitamin D cellular response to interleukin-1 cellular response to tumor necrosis factor cellular response to redox state response to dexamethasone regulation of blood vessel size cellular response to transforming growth factor beta stimulus response to fructose response to metal ion response to cytokine cellular response to heat response to statin response to drug response to amino acid response to axon injury response to corticosterone response to hyperoxia response to lipoprotein particle retina development in camera-type eye bone development cellular response to magnesium ion cellular response to cAMP cellular response to organic cyclic compound response to candesartan response to peptide response to ketone response to salt
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline