Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • ADORA2A Antibodies

          Invitrogen

          ADORA2A Polyclonal Antibody

          1 Published Figure
          9 References
          View all (21) ADORA2A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ADORA2A Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          • Published Figures (1)
          ADORA2A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ADORA2A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ADORA2A Antibody (PA1-042) in ICC/IF

          Immunofluorescent analysis of Adenosine Receptor A2a (green) showing staining in the cytoplasm of U251 cells (right) compared to a negative control without primary antibody (left). Formalin-fixed cells were permeabilized with 0.1% Triton X-100 in TBS for 5-10 minutes and blocked with 3% BSA-PBS for 30 minutes at room temperature. Cells were probed with an Adenosine Receptor A2a polyclonal antibody (Product # PA1-042) in 3% BSA-PBS at a dilution of 1:20 and incubated overnight at 4 °C in a humidified chamber. Cells were washed with PBST and... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ADORA2A Antibody in Immunocytochemistry (ICC/IF)
          ADORA2A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ADORA2A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ADORA2A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ADORA2A Antibody in Western Blot (WB)
          ADORA2A Antibody in Western Blot (WB)
          ADORA2A Polyclonal Antibody

          Product Details

          PA1-042

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 4 publications 4 publications

          Immunohistochemistry (IHC)

          -
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Dog, Human, Mouse, Rat

          Published species

          Dog, Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues E(373) S H G D M G L P D V E L L S H E L K(391) of canine A2aAR.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.69 mg/mL

          Amount

          69 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2257858

          Product Specific Information

          PA1-042 detects adenosine receptor A2a (A2aAR) in human, rat, mouse and canine tissues. This antibody does not detect other AR subtypes.

          PA1-042 has been successfully used in Western blot, ICC/IF, immunohistochemistry and immunoprecipitation procedures. By Western blot, this antibody detects an ~45 kDa protein from canine striatum representing A2aAR. Immunohistochemical staining of A2aAR in human hippocampus with PA1-042 yields a pattern consistent with plasma membrane staining.

          The PA1-042 immunogen is a synthetic peptide corresponding to residues E(373) S H G D M G L P D V E L L S H E L K(391) of canine A2aAR.

          Target Information

          ADORA2A is a protein which is one of several receptor subtypes for adenosine. The activity of the encoded protein, a G-protein coupled receptor family member, is mediated by G proteins which activate adenylyl cyclase. This protein is abundant in basal ganglia, vasculature and platelets and it is a major target of caffeine.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: A2a receptor; Adenosine A2a-receptor; Adenosine receptor A2a; adenosine receptor subtype A2a; ADORA2A; AR; hA2aR; unnamed protein product

          View more View less

          Gene Aliases: A2AAR; A2aR; AA2AR; ADENO; ADORA2; ADORA2A; Adora2l1; RDC8

          View more View less

          UniProt ID: (Human) P29274, (Dog) P11617, (Mouse) Q60613, (Rat) P30543

          View more View less

          Entrez Gene ID: (Human) 135, (Dog) 403960, (Mouse) 11540, (Rat) 25369

          View more View less

          Function(s)
          G-protein coupled adenosine receptor activity G-protein coupled receptor activity protein binding calmodulin binding lipid binding enzyme binding type 5 metabotropic glutamate receptor binding identical protein binding macromolecular complex binding alpha-actinin binding signal transducer activity protein heterodimerization activity G-protein coupled receptor
          Process(es)
          synaptic transmission, dopaminergic adenosine receptor signaling pathway response to amphetamine regulation of transcription, DNA-templated phagocytosis apoptotic process inflammatory response cellular defense response signal transduction G-protein coupled receptor signaling pathway adenylate cyclase-modulating G-protein coupled receptor signaling pathway adenylate cyclase-activating G-protein coupled receptor signaling pathway phospholipase C-activating G-protein coupled receptor signaling pathway cell-cell signaling synaptic transmission, cholinergic central nervous system development blood coagulation sensory perception locomotory behavior blood circulation negative regulation of cell proliferation response to xenobiotic stimulus positive regulation of glutamate secretion positive regulation of acetylcholine secretion, neurotransmission regulation of norepinephrine secretion response to purine-containing compound response to caffeine positive regulation of synaptic transmission, GABAergic synaptic transmission, glutamatergic positive regulation of urine volume vasodilation eating behavior negative regulation of vascular permeability response to alkaloid negative regulation of neuron apoptotic process positive regulation of circadian sleep/wake cycle, sleep negative regulation of alpha-beta T cell activation astrocyte activation neuron projection morphogenesis positive regulation of protein secretion negative regulation of inflammatory response regulation of mitochondrial membrane potential membrane depolarization regulation of calcium ion transport positive regulation of synaptic transmission, glutamatergic excitatory postsynaptic potential inhibitory postsynaptic potential prepulse inhibition positive regulation of ERK1 and ERK2 cascade apoptotic signaling pathway presynaptic modulation of chemical synaptic transmission positive regulation of long-term synaptic potentiation positive regulation of apoptotic signaling pathway negative regulation of protein kinase activity protein kinase C-activating G-protein coupled receptor signaling pathway positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway positive regulation of cAMP biosynthetic process positive regulation of renal sodium excretion negative regulation of locomotion negative regulation of cysteine-type endopeptidase activity involved in apoptotic process regulation of synaptic plasticity response to drug
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline