Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • alpha-2a Adrenergic Receptor Antibodies

          Invitrogen

          alpha-2a Adrenergic Receptor Polyclonal Antibody

          8 Published Figures
          15 References
          View all (13) alpha-2a Adrenergic Receptor antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite alpha-2a Adrenergic Receptor Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (8)
          alpha-2a Adrenergic Receptor Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          alpha-2a Adrenergic Receptor Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 9

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          alpha-2a Adrenergic Receptor Antibody (PA1-048) in WB

          Western blot analysis was performed on whole cell extracts (30 µg lysate) of PC-3 (Lane 1), HEK-293 (Lane 2), T47D (Lane 3) and HEL 92.1.7 (lane 4). The blots were probed with Anti-alpha-2a Adrenergic Receptor Rabbit Polyclonal Antibody (Product # PA1-048, 1:250-1:1000 dilution) and detected by chemiluminescence using Goat anti-Rabbit IgG (H+L) Secondary Antibody, HRP conjugate (Product # G-21234, 1:5000 di... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          alpha-2a Adrenergic Receptor Antibody in Western Blot (WB)
          alpha-2a Adrenergic Receptor Antibody in Immunohistochemistry (Frozen) (IHC (F))
          alpha-2a Adrenergic Receptor Antibody in Immunohistochemistry (IHC)
          alpha-2a Adrenergic Receptor Antibody in Immunohistochemistry (IHC)
          alpha-2a Adrenergic Receptor Antibody in Western Blot (WB)
          alpha-2a Adrenergic Receptor Antibody in Western Blot (WB)
          alpha-2a Adrenergic Receptor Antibody in Western Blot (WB)
          alpha-2a Adrenergic Receptor Antibody in Western Blot (WB)
          alpha-2a Adrenergic Receptor Antibody in Western Blot (WB)
          alpha-2a Adrenergic Receptor Polyclonal Antibody

          Product Details

          PA1-048

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500
          View 6 publications 6 publications

          Immunohistochemistry (IHC)

          -
          View 6 publications 6 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:1,000
          -

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corressponding to residues R(218) I Y Q I A K R R T R V P P S R R G(235) of the 3rd intracellular loop of human A2AAR.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.6 mg/mL

          Amount

          60 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2225243

          Product Specific Information

          PA1-048 detects alpha-2A adrenergic receptor (A2AAR) from human, rat and mouse tissues.

          PA1-048 has been successfully used in Western blot and immunohistochemistry procedures. By Western blot, this antibody detects an ~45 kDa protein representing A2AAR from mouse kidney membrane preparations.

          The PA1-048 immunogen is a synthetic peptide corressponding to residues R(218) I Y Q I A K R R T R V P P S R R G(235) of the 3rd intracellular loop of human A2AAR. This sequence is completely conserved between human, mouse, rat, and porcine A2AAR.

          Target Information

          Alpha-2-adrenergic receptors (ADRA2A) are members of the G protein-coupled receptor superfamily. They include 3 highly homologous subtypes: alpha2A, alpha2B, and alpha2C. ADRA2A mediate the catecholamine-induced inhibition of adenylate cyclase through the action of G protein. These receptors have a critical role in regulating neurotransmitter release from sympathetic nerves and from adrenergic neurons in the central nervous system.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: adrenergic receptor, alpha 2a; adrenergic, alpha-2A-, receptor; Alpha-2 adrenergic receptor subtype C10; alpha-2-adrenergic receptor, platelet type; Alpha-2A adrenergic receptor; Alpha-2A adrenoceptor; Alpha-2A adrenoreceptor; alpha-2AAR; alpha-2AAR subtype C10; Alpha-2D adrenergic receptor; alpha2A-adrenergic receptor; CA2-47; OTTHUMP00000058849; subtype alpha2-C10

          View more View less

          Gene Aliases: Adra-2; Adra-2a; ADRA2; ADRA2A; ADRA2R; ADRAR; alpha(2A)AR; alpha2-C10; alpha2A; alpha2A-AR; ALPHA2AAR; AW122659; CA2-47; FPLD8; RATRG20; RG20; ZNF32

          View more View less

          UniProt ID: (Human) P08913, (Mouse) Q01338, (Rat) P22909

          View more View less

          Entrez Gene ID: (Human) 150, (Mouse) 11551, (Rat) 25083

          View more View less

          Function(s)
          G-protein coupled receptor activity adrenergic receptor activity alpha2-adrenergic receptor activity guanyl-nucleotide exchange factor activity protein binding protein kinase binding alpha-1B adrenergic receptor binding alpha-2C adrenergic receptor binding thioesterase binding heterotrimeric G-protein binding protein homodimerization activity protein heterodimerization activity epinephrine binding norepinephrine binding G-protein coupled receptor
          Process(es)
          positive regulation of cytokine production DNA replication regulation of smooth muscle contraction signal transduction epidermal growth factor receptor signaling pathway G-protein coupled receptor signaling pathway adenylate cyclase-activating G-protein coupled receptor signaling pathway adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway Ras protein signal transduction Rho protein signal transduction female pregnancy positive regulation of cell proliferation negative regulation of norepinephrine secretion regulation of vasoconstriction actin cytoskeleton organization platelet activation positive regulation of cell migration negative regulation of epinephrine secretion cellular response to hormone stimulus vasodilation glucose homeostasis fear response positive regulation of potassium ion transport response to morphine positive regulation of MAPK cascade positive regulation of epidermal growth factor receptor signaling pathway negative regulation of calcium ion-dependent exocytosis negative regulation of insulin secretion intestinal absorption thermoception negative regulation of lipid catabolic process positive regulation of membrane protein ectodomain proteolysis positive regulation of protein kinase B signaling negative regulation of calcium ion transport negative regulation of insulin secretion involved in cellular response to glucose stimulus negative regulation of uterine smooth muscle contraction adrenergic receptor signaling pathway adenylate cyclase-activating adrenergic receptor signaling pathway adenylate cyclase-inhibiting adrenergic receptor signaling pathway phospholipase C-activating adrenergic receptor signaling pathway positive regulation of wound healing response to alcohol presynaptic modulation of chemical synaptic transmission acute inflammatory response negative regulation of adenylate cyclase activity cell-cell signaling negative regulation of cAMP biosynthetic process activation of protein kinase activity activation of protein kinase B activity epidermal growth factor-activated receptor transactivation by G-protein coupled receptor signaling pathway positive regulation of MAP kinase activity positive regulation of epidermal growth factor-activated receptor activity positive regulation of vasodilation activation of MAPK activity by adrenergic receptor signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline