Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Furin Antibodies

          Invitrogen

          Furin Polyclonal Antibody

          Advanced Verification
          31 Published Figures
          55 References
          View all (20) Furin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Furin Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          • Published Figures (31)
          • Advanced Verification (4)
          Furin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Furin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 41

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Furin Antibody (PA1-062) in ICC/IF

          Immunofluorescence analysis of Furin was performed using 70% confluent log phase BeWo cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 45 minutes at room temperature. The cells were labeled with Furin Polyclonal Antibody (Product # PA1-062) at 2 µg/mL dilution in 0.1% BSA, incubated at 4 degree celsius overnight and then labeled with Donkey anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor Plus 488 (Product # A327... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Furin Antibody in Immunocytochemistry (ICC/IF)
          Furin Antibody in Immunocytochemistry (ICC/IF)
          Furin Antibody in Immunocytochemistry (ICC/IF)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Immunohistochemistry (IHC)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Immunocytochemistry (ICC/IF)
          Furin Antibody in Immunocytochemistry (ICC/IF)
          Furin Antibody in Immunocytochemistry (ICC/IF)
          Furin Antibody in Immunocytochemistry (ICC/IF)
          Furin Antibody in Western Blot, Immunocytochemistry, Immunoprecipitation (WB, ICC/IF, IP)
          Furin Antibody in Immunocytochemistry (ICC/IF)
          Furin Antibody in Immunocytochemistry (ICC/IF)
          Furin Antibody in Immunocytochemistry (ICC/IF)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Western Blot, Immunoprecipitation (WB, IP)
          Furin Antibody in Western Blot, Immunoprecipitation (WB, IP)
          Furin Antibody in Western Blot (WB)
          Furin Antibody in Immunoprecipitation (IP)
          Furin Antibody in Immunoprecipitation (IP)
          Furin Antibody
          Furin Antibody
          Furin Antibody
          Furin Antibody
          Furin Polyclonal Antibody

          Product Details

          PA1-062

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 22 publications 22 publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunocytochemistry (ICC/IF)

          2 µg/mL
          View 24 publications 24 publications

          Immunoprecipitation (IP)

          1:100
          View 3 publications 3 publications

          Neutralization (Neu)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Dog, Human, Mouse, Rat

          Published species

          Dog, Hamster, Human, Mouse, Non-human primate, Pig, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues R(780) G E R T A F I K D Q S A L(793) of human Furin.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          2 mg/mL

          Amount

          200 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2105077

          Product Specific Information

          PA1-062 detects furin convertase from canine and mouse cells as well as transfected human furin. This antibody does not detect endogenous furin from BSC-40, HeLa, J774A.1 BPAEC, or CHO cells, or from rat skeletal muscle, spleen, kidney, ovary, testes, heart, or brain tissue.

          PA1-062 has been successfully used in Western blot, immunocytochemistry, immunoprecipitation, and immunofluorescence procedures. By Western blot, this antibody detects an ~100 kDa protein representing furin in BSC-40 cells transfected with the human furin gene. In addition, PA1-062 detects human furin transfected into 3T3 cells. Immunofluorescence staining of endogenous furin convertase in 3T3 cells with PA1-062 yields a pattern consistent with paranuclear staining.

          The PA1-062 immunogen is a synthetic peptide corresponding to residues R(780) G E R T A F I K D Q S A L(793) of human Furin. The PA1-062 immunizing peptide (Cat. # PEP-010) is available for use in neutralization and control experiments.

          Target Information

          Furin is a membrane-associated, calcium-dependent, serine protease that belongs to the subtilisin-like prohormone convertase (PC) family. Members of this family of cellular enzymes cleave most prohormones and neuropeptide precursors. Numerous other cellular proteins, some viral proteins, and bacterial toxins that are transported by the constitutive secretory pathway are also targeted for maturation by PCs. Furin and other PC family members share structural similarities which include a heterogeneous ~10 kDa amino-terminal proregion, a highly conserved ~55 kDa subtilisin-like catalytic domain, and carboxyl-terminal domain that is heterogeneous in length and sequence. These enzymes become catalytically active following proregion cleavage within the appropriate cellular compartment. Furin is the only known PC to possess a transmembrane domain. Cleavage of target proteins occurs at the carboxyl-terminus of the furin consensus sequence, RX(K/R)R. It has been shown that the acidic peptide sequence, C771PSDSEEDEG780, localizes furin to the trans-Golgi-network. Phosphorylation of serine residues within this region modulates intracellular routing of furin protein. An additional signaling domain includes the tetrapeptide sequence, Y759KGL762, which directs internalization from the cell surface.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: dibasic processing enzyme; Dibasic-processing enzyme; FES upstream region; Furin; furin, membrane associated receptor protein; PACE; Paired basic amino acid cleaving enzyme (furin); paired basic amino acid residue cleaving enzyme; Paired basic amino acid residue-cleaving enzyme; Prohormone convertase 3; proprotein convertase subtilisin/kexin 3; proprotein convertase subtilisin/kexin type 3; Proprotein convertase subtilisin/kexin type 3 (paired basic amino acid cleaving enzyme, furin, membrane associated receptor protein); proprotein convertase subtilisin/kexin type3; subtilisin pro-protein processing enzyme; subtilisin-like proprotein convertase 1; unnamed protein product

          View more View less

          Gene Aliases: 9130404I01Rik; FUR; FURIN; PACE; PCSK3; SPC1

          View more View less

          UniProt ID: (Human) P09958, (Rat) P23377, (Mouse) P23188

          View more View less

          Entrez Gene ID: (Dog) 488746, (Human) 5045, (Rat) 54281, (Mouse) 18550

          View more View less

          Function(s)
          protease binding endopeptidase activity serine-type endopeptidase activity serine-type endopeptidase inhibitor activity protein binding heparin binding peptidase activity serine-type peptidase activity hydrolase activity peptide binding metal ion binding nerve growth factor binding endopeptidase activator activity heparan sulfate binding serine protease
          Process(es)
          blastocyst formation negative regulation of inflammatory response to antigenic stimulus signal peptide processing proteolysis transforming growth factor beta receptor signaling pathway regulation of signal transduction protein processing peptide hormone processing viral life cycle viral translation viral protein processing extracellular matrix disassembly extracellular matrix organization collagen catabolic process zymogen activation regulation of cholesterol transport negative regulation of low-density lipoprotein particle receptor catabolic process nerve growth factor production negative regulation of nerve growth factor production negative regulation of transforming growth factor beta1 production secretion by cell plasma lipoprotein particle remodeling regulation of protein catabolic process peptide biosynthetic process positive regulation of viral entry into host cell viral entry into host cell positive regulation of membrane protein ectodomain proteolysis protein maturation dibasic protein processing zymogen inhibition cytokine precursor processing amyloid fibril formation aging cell proliferation negative regulation of endopeptidase activity positive regulation of cell migration positive regulation of transforming growth factor beta receptor signaling pathway regulation of low-density lipoprotein particle receptor biosynthetic process regulation of endopeptidase activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline